Gene Transcripts
- Gene Symbol
- KRT42P
- Entrez Gene ID
- 284116
- Gene Name
- keratin 42 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 17: 41626327 - 41640199 on Build GRCh38
- UniGene ID
- Hs.725790
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033415.1 |
|
3, 4 |
1027 |
|
GenBank mRNA |
AK095281.1 |
- |
2, 3 |
347 |
|
AK125965.1 |
- |
3, 4 |
1027 |
|
- Gene Symbol
- AGTPBP1
- Entrez Gene ID
- 23287
- Gene Name
- ATP/GTP binding protein 1
- Gene Aliases
- CCP1, NNA1
- Chromosome Location
- Chr. 9: 85543799 - 85742029 on Build GRCh38
- UniGene ID
- Hs.719980
- Species
- Homo sapiens
- Gene Symbol
- AIFM1
- Entrez Gene ID
- 9131
- Gene Name
- apoptosis inducing factor, mitochondria associated 1
- Gene Aliases
- AIF, CMT2D, CMTX4, COWCK, COXPD6, DFNX5, NADMR, NAMSD, PDCD8
- Chromosome Location
- Chr. X: 130129362 - 130165887 on Build GRCh38
- UniGene ID
- Hs.424932
- Species
- Homo sapiens
- Gene Symbol
- AMDHD2
- Entrez Gene ID
- 51005
- Gene Name
- amidohydrolase domain containing 2
- Gene Aliases
- CGI-14
- Chromosome Location
- Chr. 16: 2520351 - 2531414 on Build GRCh38
- UniGene ID
- Hs.740430
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209351.1 |
- |
1 |
2441 |
|
- Gene Symbol
- AURKA
- Entrez Gene ID
- 6790
- Gene Name
- aurora kinase A
- Gene Aliases
- AIK, ARK1, AURA, BTAK, PPP1R47, STK15, STK6, STK7
- Chromosome Location
- Chr. 20: 56369389 - 56392337 on Build GRCh38
- UniGene ID
- Hs.250822
- Species
- Homo sapiens
- Gene Symbol
- B3GALNT2
- Entrez Gene ID
- 148789
- Gene Name
- beta-1,3-N-acetylgalactosaminyltransferase 2
- Gene Aliases
- B3GalNAc-T2, MDDGA11
- Chromosome Location
- Chr. 1: 235440654 - 235504481 on Build GRCh38
- UniGene ID
- Hs.498143
- Species
- Homo sapiens
- Gene Symbol
- C10orf126
- Entrez Gene ID
- 283080
- Gene Name
- chromosome 10 open reading frame 126
- Gene Aliases
- bA492M23.1
- Chromosome Location
- Chr. 10: 28846408 - 28881898 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- C21orf62
- Entrez Gene ID
- 56245
- Gene Name
- chromosome 21 open reading frame 62
- Gene Aliases
- B37, C21orf120, PRED81
- Chromosome Location
- Chr. 21: 32790673 - 32813808 on Build GRCh38
- UniGene ID
- Hs.517235
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BM512397.1 |
- |
1 |
51 |
|
BU789528.1 |
- |
1 |
51 |
|
- Gene Symbol
- C3orf33
- Entrez Gene ID
- 285315
- Gene Name
- chromosome 3 open reading frame 33
- Gene Aliases
- AC3-33
- Chromosome Location
- Chr. 3: 155762612 - 155806287 on Build GRCh38
- UniGene ID
- Hs.350846
- Species
- Homo sapiens
- Gene Symbol
- CARD6
- Entrez Gene ID
- 84674
- Gene Name
- caspase recruitment domain family member 6
- Gene Aliases
- CINCIN1
- Chromosome Location
- Chr. 5: 40841190 - 40855354 on Build GRCh38
- UniGene ID
- Hs.200242
- Species
- Homo sapiens
- Gene Symbol
- CEP135
- Entrez Gene ID
- 9662
- Gene Name
- centrosomal protein 135
- Gene Aliases
- CEP4, KIAA0635, MCPH8
- Chromosome Location
- Chr. 4: 55948808 - 56033363 on Build GRCh38
- UniGene ID
- Hs.518767
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK309554.1 |
- |
5 |
1064 |
|
- Gene Symbol
- DDX46
- Entrez Gene ID
- 9879
- Gene Name
- DEAD-box helicase 46
- Gene Aliases
- PRPF5, Prp5
- Chromosome Location
- Chr. 5: 134758734 - 134831138 on Build GRCh38
- UniGene ID
- Hs.406549
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AI003503.1 |
- |
1 |
275 |
|
- Gene Symbol
- DPY19L1P1
- Entrez Gene ID
- 100129460
- Gene Name
- DPY19L1 pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 7: 32580939 - 32719168 on Build GRCh38
- UniGene ID
- Hs.633705
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_036680.1 |
|
13 |
1059 |
|
GenBank mRNA |
AK026768.1 |
- |
13 |
1062 |
|
- Gene Symbol
- GNG7
- Entrez Gene ID
- 2788
- Gene Name
- G protein subunit gamma 7
- Gene Aliases
- Chromosome Location
- Chr. 19: 2511219 - 2702748 on Build GRCh38
- UniGene ID
- Hs.515544
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK024465.1 |
- |
1 |
889 |
|
- Gene Symbol
- GPR1
- Entrez Gene ID
- 2825
- Gene Name
- G protein-coupled receptor 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 206175316 - 206218047 on Build GRCh38
- UniGene ID
- Hs.184907
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CA446989.1 |
- |
1 |
249 |
|
- Gene Symbol
- IPP
- Entrez Gene ID
- 3652
- Gene Name
- intracisternal A particle-promoted polypeptide
- Gene Aliases
- KLHL27
- Chromosome Location
- Chr. 1: 45694324 - 45750813 on Build GRCh38
- UniGene ID
- Hs.744252
- Species
- Homo sapiens
- Gene Symbol
- LIN52
- Entrez Gene ID
- 91750
- Gene Name
- lin-52 DREAM MuvB core complex component
- Gene Aliases
- C14orf46, c14_5549
- Chromosome Location
- Chr. 14: 74084831 - 74201255 on Build GRCh38
- UniGene ID
- Hs.612866
- Species
- Homo sapiens
- Gene Symbol
- LINC01470
- Entrez Gene ID
- 101927134
- Gene Name
- long intergenic non-protein coding RNA 1470
- Gene Aliases
- Chromosome Location
- Chr. 5: 152618965 - 152972349 on Build GRCh38
- UniGene ID
- Hs.531423
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092848.1 |
- |
3 |
285 |
|
- Gene Symbol
- LOC101927189
- Entrez Gene ID
- 101927189
- Gene Name
- uncharacterized LOC101927189
- Gene Aliases
- Chromosome Location
- Chr. 6: 53929982 - 53997667 on Build GRCh38
- UniGene ID
- Hs.453592
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_125842.1 |
|
3 |
422 |
|
GenBank mRNA |
AK056584.1 |
- |
3 |
223 |
|
BX640790.1 |
- |
3 |
361 |
|
- Gene Symbol
- LOC105371775
- Entrez Gene ID
- 105371775
- Gene Name
- uncharacterized LOC105371775
- Gene Aliases
- Chromosome Location
- Chr. 17: 40671605 - 40697840 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL110162.1 |
- |
1 |
112 |
|
- Gene Symbol
- LOC105372966
- Entrez Gene ID
- 105372966
- Gene Name
- uncharacterized LOC105372966
- Gene Aliases
- Chromosome Location
- Chr. 22: 25476218 - 25479939 on Build GRCh38
- UniGene ID
- Hs.667879
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124562.1 |
- |
1 |
24 |
|
- Gene Symbol
- LRIG2
- Entrez Gene ID
- 9860
- Gene Name
- leucine rich repeats and immunoglobulin like domains 2
- Gene Aliases
- LIG-2, LIG2, UFS2
- Chromosome Location
- Chr. 1: 113073170 - 113132260 on Build GRCh38
- UniGene ID
- Hs.448972
- Species
- Homo sapiens
- Gene Symbol
- MBD3
- Entrez Gene ID
- 53615
- Gene Name
- methyl-CpG binding domain protein 3
- Gene Aliases
- Chromosome Location
- Chr. 19: 1576671 - 1592761 on Build GRCh38
- UniGene ID
- Hs.178728
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092508.1 |
- |
1 |
1214 |
|
AL390153.1 |
- |
1 |
784 |
|
- Gene Symbol
- MELK
- Entrez Gene ID
- 9833
- Gene Name
- maternal embryonic leucine zipper kinase
- Gene Aliases
- HPK38
- Chromosome Location
- Chr. 9: 36572862 - 36677683 on Build GRCh38
- UniGene ID
- Hs.184339
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046337.1 |
|
7 |
698 |
|
GenBank mRNA |
AK296820.1 |
- |
7 |
697 |
|
- Gene Symbol
- MSH5
- Entrez Gene ID
- 4439
- Gene Name
- mutS homolog 5
- Gene Aliases
- G7, MUTSH5, NG23
- Chromosome Location
- Chr. 6: 31739948 - 31762678 on Build GRCh38
- UniGene ID
- Hs.647011
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209886.1 |
- |
2 |
2191 |
|
- Gene Symbol
- NDST3
- Entrez Gene ID
- 9348
- Gene Name
- N-deacetylase and N-sulfotransferase 3
- Gene Aliases
- HSST3
- Chromosome Location
- Chr. 4: 118033596 - 118258945 on Build GRCh38
- UniGene ID
- Hs.480596
- Species
- Homo sapiens
- Gene Symbol
- NFKBID
- Entrez Gene ID
- 84807
- Gene Name
- NFKB inhibitor delta
- Gene Aliases
- IkappaBNS, TA-NFKBH
- Chromosome Location
- Chr. 19: 35887630 - 35902301 on Build GRCh38
- UniGene ID
- Hs.466531
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126825.1 |
- |
4 |
1406 |
|
- Gene Symbol
- NSG1
- Entrez Gene ID
- 27065
- Gene Name
- neuron specific gene family member 1
- Gene Aliases
- D4S234, D4S234E, NEEP21, P21
- Chromosome Location
- Chr. 4: 4380988 - 4419058 on Build GRCh38
- UniGene ID
- Hs.518595
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094573.1 |
- |
2 |
129 |
|
- Gene Symbol
- PDHA1
- Entrez Gene ID
- 5160
- Gene Name
- pyruvate dehydrogenase (lipoamide) alpha 1
- Gene Aliases
- PDHA, PDHAD, PDHCE1A, PHE1A
- Chromosome Location
- Chr. X: 19343893 - 19361707 on Build GRCh38
- UniGene ID
- Hs.530331
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BU170389.1 |
- |
1 |
782 |
|
- Gene Symbol
- PIGQ
- Entrez Gene ID
- 9091
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis class Q
- Gene Aliases
- GPI1, c407A10.1
- Chromosome Location
- Chr. 16: 569968 - 584136 on Build GRCh38
- UniGene ID
- Hs.741878
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092073.1 |
- |
1 |
63 |
|
- Gene Symbol
- PP12708
- Entrez Gene ID
- 100130609
- Gene Name
- uncharacterized LOC100130609
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.684470
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF318327.1 |
- |
2 |
1662 |
|
- Gene Symbol
- PRKAA1
- Entrez Gene ID
- 5562
- Gene Name
- protein kinase AMP-activated catalytic subunit alpha 1
- Gene Aliases
- AMPK, AMPKa1
- Chromosome Location
- Chr. 5: 40759379 - 40798195 on Build GRCh38
- UniGene ID
- Hs.43322
- Species
- Homo sapiens
- Gene Symbol
- RAC1
- Entrez Gene ID
- 5879
- Gene Name
- ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1)
- Gene Aliases
- MIG5, Rac-1, TC-25, p21-Rac1
- Chromosome Location
- Chr. 7: 6374495 - 6403967 on Build GRCh38
- UniGene ID
- Hs.413812
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209410.1 |
- |
3 |
3105 |
|
- Gene Symbol
- RARS2
- Entrez Gene ID
- 57038
- Gene Name
- arginyl-tRNA synthetase 2, mitochondrial
- Gene Aliases
- ArgRS, DALRD2, PCH6, PRO1992, RARSL
- Chromosome Location
- Chr. 6: 87513744 - 87590032 on Build GRCh38
- UniGene ID
- Hs.485910
- Species
- Homo sapiens
- Gene Symbol
- RNF212
- Entrez Gene ID
- 285498
- Gene Name
- ring finger protein 212
- Gene Aliases
- ZHP3
- Chromosome Location
- Chr. 4: 1056248 - 1113794 on Build GRCh38
- UniGene ID
- Hs.248290
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124578.1 |
- |
1 |
2893 |
|
- Gene Symbol
- RPS6KA3
- Entrez Gene ID
- 6197
- Gene Name
- ribosomal protein S6 kinase A3
- Gene Aliases
- CLS, HU-3, ISPK-1, MAPKAPK1B, MRX19, RSK, RSK2, S6K-alpha3, p90-RSK2, pp90RSK2
- Chromosome Location
- Chr. X: 20149911 - 20267514 on Build GRCh38
- UniGene ID
- Hs.445387
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CK905714.1 |
- |
1 |
57 |
|
- Gene Symbol
- SERF2-C15ORF63
- Entrez Gene ID
- 100529067
- Gene Name
- SERF2-C15orf63 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 15: 43791976 - 43802589 on Build GRCh38
- UniGene ID
- Hs.730672
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF370428.1 |
- |
1 |
1306 |
|
- Gene Symbol
- SLC13A5
- Entrez Gene ID
- 284111
- Gene Name
- solute carrier family 13 member 5
- Gene Aliases
- EIEE25, NACT, mIndy
- Chromosome Location
- Chr. 17: 6684713 - 6713421 on Build GRCh38
- UniGene ID
- Hs.399496
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124317.1 |
- |
1 |
623 |
|
BX647354.1 |
- |
1 |
1373 |
|
- Gene Symbol
- SLC27A5
- Entrez Gene ID
- 10998
- Gene Name
- solute carrier family 27 member 5
- Gene Aliases
- ACSB, ACSVL6, BACS, BAL, FACVL3, FATP-5, FATP5, VLACSR, VLCS-H2, VLCSH2
- Chromosome Location
- Chr. 19: 58498333 - 58512065 on Build GRCh38
- UniGene ID
- Hs.292177
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK026640.1 |
- |
4 |
502 |
|
- Gene Symbol
- SOCS7
- Entrez Gene ID
- 30837
- Gene Name
- suppressor of cytokine signaling 7
- Gene Aliases
- NAP4, NCKAP4
- Chromosome Location
- Chr. 17: 38351876 - 38405593 on Build GRCh38
- UniGene ID
- Hs.514132
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC121030.1 |
- |
8 |
1721 |
|
- Gene Symbol
- SSBP1
- Entrez Gene ID
- 6742
- Gene Name
- single stranded DNA binding protein 1
- Gene Aliases
- Mt-SSB, SOSS-B1, SSBP, mtSSB
- Chromosome Location
- Chr. 7: 141738309 - 141750488 on Build GRCh38
- UniGene ID
- Hs.490394
- Species
- Homo sapiens
- Gene Symbol
- STAT2
- Entrez Gene ID
- 6773
- Gene Name
- signal transducer and activator of transcription 2
- Gene Aliases
- IMD44, ISGF-3, P113, STAT113
- Chromosome Location
- Chr. 12: 56341597 - 56360253 on Build GRCh38
- UniGene ID
- Hs.530595
- Species
- Homo sapiens
- Gene Symbol
- TMEM131
- Entrez Gene ID
- 23505
- Gene Name
- transmembrane protein 131
- Gene Aliases
- CC28, PRO1048, RW1, YR-23
- Chromosome Location
- Chr. 2: 97756336 - 97996182 on Build GRCh38
- UniGene ID
- Hs.469376
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX647728.1 |
- |
1 |
2486 |
|
- Gene Symbol
- TNPO2
- Entrez Gene ID
- 30000
- Gene Name
- transportin 2
- Gene Aliases
- IPO3, KPNB2B, TRN2
- Chromosome Location
- Chr. 19: 12699194 - 12723996 on Build GRCh38
- UniGene ID
- Hs.416049
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK022217.1 |
- |
1 |
157 |
|
- Gene Symbol
- TSPAN5
- Entrez Gene ID
- 10098
- Gene Name
- tetraspanin 5
- Gene Aliases
- NET-4, NET4, TM4SF9, TSPAN-5
- Chromosome Location
- Chr. 4: 98470367 - 98658661 on Build GRCh38
- UniGene ID
- Hs.118118
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126702.1 |
- |
1 |
451 |
|
- Gene Symbol
- TUBGCP5
- Entrez Gene ID
- 114791
- Gene Name
- tubulin gamma complex associated protein 5
- Gene Aliases
- GCP5
- Chromosome Location
- Chr. 15: 22982977 - 23039673 on Build GRCh38
- UniGene ID
- Hs.533898
- Species
- Homo sapiens
- Gene Symbol
- VCPKMT
- Entrez Gene ID
- 79609
- Gene Name
- valosin containing protein lysine methyltransferase
- Gene Aliases
- C14orf138, METTL21D, VCP-KMT
- Chromosome Location
- Chr. 14: 50100456 - 50118343 on Build GRCh38
- UniGene ID
- Hs.558541
- Species
- Homo sapiens
- Gene Symbol
- ZNF503-AS1
- Entrez Gene ID
- 253264
- Gene Name
- ZNF503 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 10: 75296383 - 75361678 on Build GRCh38
- UniGene ID
- Hs.662111
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038226.1 |
|
2 |
302 |
|
GenBank mRNA |
CN360505.1 |
- |
2 |
188 |
|
- Gene Symbol
- ZNF681
- Entrez Gene ID
- 148213
- Gene Name
- zinc finger protein 681
- Gene Aliases
- Chromosome Location
- Chr. 19: 23739195 - 23758891 on Build GRCh38
- UniGene ID
- Hs.399952
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138286.2 |
NP_612143.2
|
4 |
3236 |
|
- Gene Symbol
- ZNF75A
- Entrez Gene ID
- 7627
- Gene Name
- zinc finger protein 75a
- Gene Aliases
- Chromosome Location
- Chr. 16: 3305406 - 3322999 on Build GRCh38
- UniGene ID
- Hs.513292
- Species
- Homo sapiens
- Gene Symbol
- ZNF761
- Entrez Gene ID
- 388561
- Gene Name
- zinc finger protein 761
- Gene Aliases
- ZNF468
- Chromosome Location
- Chr. 19: 53431974 - 53458261 on Build GRCh38
- UniGene ID
- Hs.433293
- Species
- Homo sapiens
View Full Product Details