Gene Transcripts
- Gene Symbol
- C12orf50
- Entrez Gene ID
- 160419
- Gene Name
- chromosome 12 open reading frame 50
- Gene Aliases
- Chromosome Location
- Chr. 12: 87980039 - 88029399 on Build GRCh38
- UniGene ID
- Hs.112930
- Species
- Homo sapiens
- Gene Symbol
- ABCC9
- Entrez Gene ID
- 10060
- Gene Name
- ATP binding cassette subfamily C member 9
- Gene Aliases
- ABC37, ATFB12, CANTU, CMD1O, SUR2
- Chromosome Location
- Chr. 12: 21797389 - 21941869 on Build GRCh38
- UniGene ID
- Hs.732701
- Species
- Homo sapiens
- Gene Symbol
- C5orf42
- Entrez Gene ID
- 65250
- Gene Name
- chromosome 5 open reading frame 42
- Gene Aliases
- Hug, JBTS17, OFD6
- Chromosome Location
- Chr. 5: 37063928 - 37249428 on Build GRCh38
- UniGene ID
- Hs.586199
- Species
- Homo sapiens
- Gene Symbol
- CACHD1
- Entrez Gene ID
- 57685
- Gene Name
- cache domain containing 1
- Gene Aliases
- Chromosome Location
- Chr. 1: 64470793 - 64693058 on Build GRCh38
- UniGene ID
- Hs.443891
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK131297.1 |
- |
3 |
984 |
|
- Gene Symbol
- CFLAR
- Entrez Gene ID
- 8837
- Gene Name
- CASP8 and FADD like apoptosis regulator
- Gene Aliases
- CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS
- Chromosome Location
- Chr. 2: 201116015 - 201172688 on Build GRCh38
- UniGene ID
- Hs.390736
- Species
- Homo sapiens
- Gene Symbol
- CYSLTR1
- Entrez Gene ID
- 10800
- Gene Name
- cysteinyl leukotriene receptor 1
- Gene Aliases
- CYSLT1, CYSLT1R, CYSLTR, HMTMF81
- Chromosome Location
- Chr. X: 78271468 - 78327691 on Build GRCh38
- UniGene ID
- Hs.733809
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK023929.1 |
- |
2 |
2009 |
|
- Gene Symbol
- DDHD1
- Entrez Gene ID
- 80821
- Gene Name
- DDHD domain containing 1
- Gene Aliases
- PA-PLA1, PAPLA1, SPG28
- Chromosome Location
- Chr. 14: 53036740 - 53153328 on Build GRCh38
- UniGene ID
- Hs.125525
- Species
- Homo sapiens
- Gene Symbol
- DHRS4L2
- Entrez Gene ID
- 317749
- Gene Name
- dehydrogenase/reductase 4 like 2
- Gene Aliases
- SDR25C3
- Chromosome Location
- Chr. 14: 23969874 - 24006408 on Build GRCh38
- UniGene ID
- Hs.743442
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC041992.1 |
- |
2 |
1130 |
|
- Gene Symbol
- ESR1
- Entrez Gene ID
- 2099
- Gene Name
- estrogen receptor 1
- Gene Aliases
- ER, ESR, ESRA, ESTRR, Era, NR3A1
- Chromosome Location
- Chr. 6: 151654148 - 152103274 on Build GRCh38
- UniGene ID
- Hs.208124
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M69297.1 |
- |
3 |
1512 |
|
- Gene Symbol
- FLJ22447
- Entrez Gene ID
- 400221
- Gene Name
- uncharacterized LOC400221
- Gene Aliases
- Chromosome Location
- Chr. 14: 61570540 - 61654713 on Build GRCh38
- UniGene ID
- Hs.344000
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK026100.1 |
- |
5 |
845 |
|
- Gene Symbol
- FSIP2
- Entrez Gene ID
- 401024
- Gene Name
- fibrous sheath interacting protein 2
- Gene Aliases
- Chromosome Location
- Chr. 2: 185738895 - 185833289 on Build GRCh38
- UniGene ID
- Hs.98025
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092099.1 |
- |
8 |
2700 |
|
- Gene Symbol
- HELLPAR
- Entrez Gene ID
- 101101692
- Gene Name
- HELLP associated long non-coding RNA
- Gene Aliases
- LINC-HELLP, lncHELLP
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.160562
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
JX088243.1 |
- |
1 |
57826 |
|
- Gene Symbol
- KCNQ1OT1
- Entrez Gene ID
- 10984
- Gene Name
- KCNQ1 opposite strand/antisense transcript 1 (non-protein coding)
- Gene Aliases
- KCNQ1-AS2, KCNQ10T1, Kncq1, KvDMR1, KvLQT1-AS, LIT1, NCRNA00012
- Chromosome Location
- Chr. 11: 2608328 - 2699998 on Build GRCh38
- UniGene ID
- Hs.604823
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002728.3 |
|
1 |
65010 |
|
- Gene Symbol
- LCLAT1
- Entrez Gene ID
- 253558
- Gene Name
- lysocardiolipin acyltransferase 1
- Gene Aliases
- 1AGPAT8, AGPAT8, ALCAT1, HSRG1849, LYCAT, UNQ1849
- Chromosome Location
- Chr. 2: 30447226 - 30644225 on Build GRCh38
- UniGene ID
- Hs.468048
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095284.1 |
- |
7 |
1872 |
|
- Gene Symbol
- LINC00504
- Entrez Gene ID
- 201853
- Gene Name
- long intergenic non-protein coding RNA 504
- Gene Aliases
- Chromosome Location
- Chr. 4: 14470465 - 14888169 on Build GRCh38
- UniGene ID
- Hs.551051
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126435.1 |
|
7 |
3136 |
|
- Gene Symbol
- LOC100131257
- Entrez Gene ID
- 100131257
- Gene Name
- zinc finger protein 655 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 7: 7075770 - 7096786 on Build GRCh38
- UniGene ID
- Hs.551110
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034022.1 |
|
1 |
12540 |
|
- Gene Symbol
- LOC101927374
- Entrez Gene ID
- 101927374
- Gene Name
- uncharacterized LOC101927374
- Gene Aliases
- Chromosome Location
- Chr. 3: 79411727 - 79435227 on Build GRCh38
- UniGene ID
- Hs.570644
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110133.1 |
|
3 |
1449 |
|
GenBank mRNA |
AK094694.1 |
- |
3 |
1449 |
|
- Gene Symbol
- LOC101929567
- Entrez Gene ID
- 101929567
- Gene Name
- uncharacterized LOC101929567
- Gene Aliases
- Chromosome Location
- Chr. 2: 8559833 - 8583792 on Build GRCh38
- UniGene ID
- Hs.634706
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110257.1 |
|
6 |
6790 |
|
GenBank mRNA |
BX641130.1 |
- |
6 |
6797 |
|
- Gene Symbol
- LOC105379526
- Entrez Gene ID
- 105379526
- Gene Name
- uncharacterized LOC105379526
- Gene Aliases
- Chromosome Location
- Chr. 9|NT_187374: 163667 - 169753 on Build GRCh38
- UniGene ID
- Hs.591892
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X97876.1 |
- |
2 |
464 |
|
- Gene Symbol
- LOC286437
- Entrez Gene ID
- 286437
- Gene Name
- uncharacterized LOC286437
- Gene Aliases
- Chromosome Location
- Chr. X: 104122305 - 104124632 on Build GRCh38
- UniGene ID
- Hs.656786
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_039980.1 |
|
1 |
263 |
|
GenBank mRNA |
AK093652.1 |
- |
1 |
263 |
|
- Gene Symbol
- LOC646588
- Entrez Gene ID
- 646588
- Gene Name
- uncharacterized LOC646588
- Gene Aliases
- Chromosome Location
- Chr. 7: 25593301 - 25790051 on Build GRCh38
- UniGene ID
- Hs.72068
- Species
- Homo sapiens
- Gene Symbol
- NME7
- Entrez Gene ID
- 29922
- Gene Name
- NME/NM23 family member 7
- Gene Aliases
- CFAP67, MN23H7, NDK 7, NDK7, nm23-H7
- Chromosome Location
- Chr. 1: 169132385 - 169367963 on Build GRCh38
- UniGene ID
- Hs.706952
- Species
- Homo sapiens
- Gene Symbol
- SHPRH
- Entrez Gene ID
- 257218
- Gene Name
- SNF2 histone linker PHD RING helicase
- Gene Aliases
- bA545I5.2
- Chromosome Location
- Chr. 6: 145857838 - 145964436 on Build GRCh38
- UniGene ID
- Hs.723297
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB095943.1 |
- |
24 |
6348 |
|
- Gene Symbol
- WDPCP
- Entrez Gene ID
- 51057
- Gene Name
- WD repeat containing planar cell polarity effector
- Gene Aliases
- BBS15, C2orf86, CHDTHP, FRITZ, FRTZ
- Chromosome Location
- Chr. 2: 63120755 - 63840825 on Build GRCh38
- UniGene ID
- Hs.414952
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX538331.1 |
- |
14 |
2653 |
|
View Full Product Details