Gene Transcripts
- Gene Symbol
- ALOX5
- Entrez Gene ID
- 240
- Gene Name
- arachidonate 5-lipoxygenase
- Gene Aliases
- 5-LO, 5-LOX, 5LPG, LOG5
- Chromosome Location
- Chr. 10: 45374166 - 45446121 on Build GRCh38
- UniGene ID
- Hs.89499
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB208946.1 |
- |
3 |
559 |
|
- Gene Symbol
- ADAMTSL3
- Entrez Gene ID
- 57188
- Gene Name
- ADAMTS like 3
- Gene Aliases
- ADAMTSL-3
- Chromosome Location
- Chr. 15: 83654086 - 84039842 on Build GRCh38
- UniGene ID
- Hs.459162
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092324.1 |
- |
9 |
2071 |
|
- Gene Symbol
- KCNQ1OT1
- Entrez Gene ID
- 10984
- Gene Name
- KCNQ1 opposite strand/antisense transcript 1 (non-protein coding)
- Gene Aliases
- KCNQ1-AS2, KCNQ10T1, Kncq1, KvDMR1, KvLQT1-AS, LIT1, NCRNA00012
- Chromosome Location
- Chr. 11: 2608328 - 2699998 on Build GRCh38
- UniGene ID
- Hs.604823
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002728.3 |
|
1 |
61580 |
|
- Gene Symbol
- KIAA1328
- Entrez Gene ID
- 57536
- Gene Name
- KIAA1328
- Gene Aliases
- Chromosome Location
- Chr. 18: 36828929 - 37232127 on Build GRCh38
- UniGene ID
- Hs.4045
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC008670.1 |
- |
2 |
1180 |
|
- Gene Symbol
- LOC101926889
- Entrez Gene ID
- 101926889
- Gene Name
- uncharacterized LOC101926889
- Gene Aliases
- Chromosome Location
- Chr. 20: 25184672 - 25196892 on Build GRCh38
- UniGene ID
- Hs.585997
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_109994.1 |
|
6 |
1775 |
|
GenBank mRNA |
AK092658.1 |
- |
6 |
1758 |
|
- Gene Symbol
- MIPOL1
- Entrez Gene ID
- 145282
- Gene Name
- mirror-image polydactyly 1
- Gene Aliases
- CCDC193
- Chromosome Location
- Chr. 14: 37197888 - 37579207 on Build GRCh38
- UniGene ID
- Hs.660396
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX647795.1 |
- |
10 |
3169 |
|
- Gene Symbol
- NME7
- Entrez Gene ID
- 29922
- Gene Name
- NME/NM23 family member 7
- Gene Aliases
- CFAP67, MN23H7, NDK 7, NDK7, nm23-H7
- Chromosome Location
- Chr. 1: 169132385 - 169367963 on Build GRCh38
- UniGene ID
- Hs.706952
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209049.1 |
- |
7 |
4429 |
|
- Gene Symbol
- SMC5-AS1
- Entrez Gene ID
- 100507299
- Gene Name
- SMC5 antisense RNA 1 (head to head)
- Gene Aliases
- MAMDC2-AS2
- Chromosome Location
- Chr. 9: 70216059 - 70258874 on Build GRCh38
- UniGene ID
- Hs.556816
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CR936772.1 |
- |
3 |
3368 |
|
View Full Product Details