Gene Transcripts
- Gene Symbol
- ICA1L
- Entrez Gene ID
- 130026
- Gene Name
- islet cell autoantigen 1 like
- Gene Aliases
- ALS2CR14, ALS2CR15
- Chromosome Location
- Chr. 2: 202773150 - 202871985 on Build GRCh38
- UniGene ID
- Hs.516629
- Species
- Homo sapiens
- Gene Symbol
- ADORA2B
- Entrez Gene ID
- 136
- Gene Name
- adenosine A2b receptor
- Gene Aliases
- ADORA2
- Chromosome Location
- Chr. 17: 15927781 - 15975896 on Build GRCh38
- UniGene ID
- Hs.167046
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000676.2 |
NP_000667.1
|
2 |
1846 |
|
GenBank mRNA |
M97759.1 |
- |
2 |
1648 |
|
- Gene Symbol
- ARID4B
- Entrez Gene ID
- 51742
- Gene Name
- AT-rich interaction domain 4B
- Gene Aliases
- BCAA, BRCAA1, RBBP1L1, RBP1L1, SAP180
- Chromosome Location
- Chr. 1: 235166895 - 235328240 on Build GRCh38
- UniGene ID
- Hs.575782
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057552.1 |
- |
11 |
1744 |
|
- Gene Symbol
- BRD1
- Entrez Gene ID
- 23774
- Gene Name
- bromodomain containing 1
- Gene Aliases
- BRL, BRPF1, BRPF2
- Chromosome Location
- Chr. 22: 49773278 - 49827927 on Build GRCh38
- UniGene ID
- Hs.127950
- Species
- Homo sapiens
- Gene Symbol
- C6orf163
- Entrez Gene ID
- 206412
- Gene Name
- chromosome 6 open reading frame 163
- Gene Aliases
- Chromosome Location
- Chr. 6: 87343080 - 87365553 on Build GRCh38
- UniGene ID
- Hs.646309
- Species
- Homo sapiens
- Gene Symbol
- CHP1
- Entrez Gene ID
- 11261
- Gene Name
- calcineurin like EF-hand protein 1
- Gene Aliases
- CHP, SLC9A1BP, Sid470p, p22, p24
- Chromosome Location
- Chr. 15: 41231149 - 41281887 on Build GRCh38
- UniGene ID
- Hs.406234
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX391712.2 |
- |
1 |
187 |
|
- Gene Symbol
- COA7
- Entrez Gene ID
- 65260
- Gene Name
- cytochrome c oxidase assembly factor 7 (putative)
- Gene Aliases
- C1orf163, RESA1, SELRC1
- Chromosome Location
- Chr. 1: 52686342 - 52698366 on Build GRCh38
- UniGene ID
- Hs.349905
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK023237.1 |
- |
1 |
1943 |
|
- Gene Symbol
- DENND5B
- Entrez Gene ID
- 160518
- Gene Name
- DENN domain containing 5B
- Gene Aliases
- Chromosome Location
- Chr. 12: 31382223 - 31591018 on Build GRCh38
- UniGene ID
- Hs.118166
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX537924.1 |
- |
14 |
3474 |
|
- Gene Symbol
- DNAJC7
- Entrez Gene ID
- 7266
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C7
- Gene Aliases
- DJ11, DJC7, TPR2, TTC2
- Chromosome Location
- Chr. 17: 41976421 - 42021371 on Build GRCh38
- UniGene ID
- Hs.500156
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK025003.1 |
- |
1 |
1527 |
|
- Gene Symbol
- DPY19L4
- Entrez Gene ID
- 286148
- Gene Name
- dpy-19 like 4 (C. elegans)
- Gene Aliases
- Chromosome Location
- Chr. 8: 94719708 - 94793848 on Build GRCh38
- UniGene ID
- Hs.567828
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC050303.1 |
- |
1 |
2242 |
|
- Gene Symbol
- ELMO1
- Entrez Gene ID
- 9844
- Gene Name
- engulfment and cell motility 1
- Gene Aliases
- CED-12, CED12, ELMO-1
- Chromosome Location
- Chr. 7: 36852906 - 37449409 on Build GRCh38
- UniGene ID
- Hs.434989
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK001084.1 |
- |
5 |
2652 |
|
- Gene Symbol
- ELMOD3
- Entrez Gene ID
- 84173
- Gene Name
- ELMO domain containing 3
- Gene Aliases
- DFNB88, LST3, RBED1, RBM29
- Chromosome Location
- Chr. 2: 85354394 - 85391752 on Build GRCh38
- UniGene ID
- Hs.269990
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC054504.1 |
- |
2 |
1672 |
|
- Gene Symbol
- FAM98B
- Entrez Gene ID
- 283742
- Gene Name
- family with sequence similarity 98 member B
- Gene Aliases
- Chromosome Location
- Chr. 15: 38454127 - 38487710 on Build GRCh38
- UniGene ID
- Hs.625289
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095745.1 |
- |
7 |
2894 |
|
- Gene Symbol
- FN1
- Entrez Gene ID
- 2335
- Gene Name
- fibronectin 1
- Gene Aliases
- CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
- Chromosome Location
- Chr. 2: 215360440 - 215436167 on Build GRCh38
- UniGene ID
- Hs.203717
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX641150.1 |
- |
7 |
4143 |
|
- Gene Symbol
- GALM
- Entrez Gene ID
- 130589
- Gene Name
- galactose mutarotase
- Gene Aliases
- BLOCK25, GLAT, HEL-S-63p, IBD1
- Chromosome Location
- Chr. 2: 38665910 - 38734767 on Build GRCh38
- UniGene ID
- Hs.435012
- Species
- Homo sapiens
- Gene Symbol
- IDI2
- Entrez Gene ID
- 91734
- Gene Name
- isopentenyl-diphosphate delta isomerase 2
- Gene Aliases
- IPPI2
- Chromosome Location
- Chr. 10: 1018907 - 1025859 on Build GRCh38
- UniGene ID
- Hs.591325
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC040651.1 |
- |
1 |
404 |
|
- Gene Symbol
- KIZ-AS1
- Entrez Gene ID
- 101929591
- Gene Name
- KIZ antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 20: 21194404 - 21218289 on Build GRCh38
- UniGene ID
- Hs.656066
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042893.1 |
- |
4 |
796 |
|
- Gene Symbol
- LOC100130987
- Entrez Gene ID
- 100130987
- Gene Name
- uncharacterized LOC100130987
- Gene Aliases
- Chromosome Location
- Chr. 11: 67317839 - 67391687 on Build GRCh38
- UniGene ID
- Hs.664585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096155.1 |
- |
4 |
2225 |
|
- Gene Symbol
- METTL2A
- Entrez Gene ID
- 339175
- Gene Name
- methyltransferase like 2A
- Gene Aliases
- METTL2
- Chromosome Location
- Chr. 17: 62423885 - 62450093 on Build GRCh38
- UniGene ID
- Hs.381204
- Species
- Homo sapiens
- Gene Symbol
- MLX
- Entrez Gene ID
- 6945
- Gene Name
- MLX, MAX dimerization protein
- Gene Aliases
- MAD7, MXD7, TCFL4, bHLHd13
- Chromosome Location
- Chr. 17: 42567060 - 42573203 on Build GRCh38
- UniGene ID
- Hs.383019
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209235.1 |
- |
|
1654 |
|
- Gene Symbol
- MOB3B
- Entrez Gene ID
- 79817
- Gene Name
- MOB kinase activator 3B
- Gene Aliases
- C9orf35, MOB1D, MOBKL2B
- Chromosome Location
- Chr. 9: 27325209 - 27529852 on Build GRCh38
- UniGene ID
- Hs.369022
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK022201.1 |
- |
1 |
2743 |
|
- Gene Symbol
- MOCS3
- Entrez Gene ID
- 27304
- Gene Name
- molybdenum cofactor synthesis 3
- Gene Aliases
- UBA4
- Chromosome Location
- Chr. 20: 50958814 - 50961863 on Build GRCh38
- UniGene ID
- Hs.159410
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
HY215239.1 |
- |
1 |
113 |
|
- Gene Symbol
- PHLDB1
- Entrez Gene ID
- 23187
- Gene Name
- pleckstrin homology like domain family B member 1
- Gene Aliases
- LL5A
- Chromosome Location
- Chr. 11: 118606296 - 118658038 on Build GRCh38
- UniGene ID
- Hs.504062
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021690.1 |
- |
2 |
1163 |
|
- Gene Symbol
- PKHD1L1
- Entrez Gene ID
- 93035
- Gene Name
- polycystic kidney and hepatic disease 1 (autosomal recessive)-like 1
- Gene Aliases
- PKHDL1
- Chromosome Location
- Chr. 8: 109362477 - 109536809 on Build GRCh38
- UniGene ID
- Hs.170128
- Species
- Homo sapiens
- Gene Symbol
- PLEKHG4B
- Entrez Gene ID
- 153478
- Gene Name
- pleckstrin homology and RhoGEF domain containing G4B
- Gene Aliases
- Chromosome Location
- Chr. 5: 54065 - 189972 on Build GRCh38
- UniGene ID
- Hs.535800
- Species
- Homo sapiens
- Gene Symbol
- PSMA2
- Entrez Gene ID
- 5683
- Gene Name
- proteasome subunit alpha 2
- Gene Aliases
- HC3, MU, PMSA2, PSC2
- Chromosome Location
- Chr. 7: 42916861 - 42932206 on Build GRCh38
- UniGene ID
- Hs.333786
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX641097.1 |
- |
5 |
4057 |
|
- Gene Symbol
- RAD9A
- Entrez Gene ID
- 5883
- Gene Name
- RAD9 checkpoint clamp component A
- Gene Aliases
- RAD9
- Chromosome Location
- Chr. 11: 67391952 - 67398412 on Build GRCh38
- UniGene ID
- Hs.655354
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK310213.1 |
- |
1 |
181 |
|
- Gene Symbol
- RPS7
- Entrez Gene ID
- 6201
- Gene Name
- ribosomal protein S7
- Gene Aliases
- DBA8, S7
- Chromosome Location
- Chr. 2: 3575263 - 3580919 on Build GRCh38
- UniGene ID
- Hs.534346
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209386.1 |
- |
2 |
1627 |
|
- Gene Symbol
- SIK2
- Entrez Gene ID
- 23235
- Gene Name
- salt inducible kinase 2
- Gene Aliases
- LOH11CR1I, QIK, SIK-2, SNF1LK2
- Chromosome Location
- Chr. 11: 111602391 - 111726917 on Build GRCh38
- UniGene ID
- Hs.269128
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096463.1 |
- |
1 |
2535 |
|
- Gene Symbol
- SLC12A6
- Entrez Gene ID
- 9990
- Gene Name
- solute carrier family 12 member 6
- Gene Aliases
- ACCPN, KCC3, KCC3A, KCC3B
- Chromosome Location
- Chr. 15: 34229996 - 34338064 on Build GRCh38
- UniGene ID
- Hs.510939
- Species
- Homo sapiens
- Gene Symbol
- SPAG16
- Entrez Gene ID
- 79582
- Gene Name
- sperm associated antigen 16
- Gene Aliases
- PF20, WDR29
- Chromosome Location
- Chr. 2: 213284369 - 214410501 on Build GRCh38
- UniGene ID
- Hs.743365
- Species
- Homo sapiens
- Gene Symbol
- SPRYD7
- Entrez Gene ID
- 57213
- Gene Name
- SPRY domain containing 7
- Gene Aliases
- C13orf1, CLLD6
- Chromosome Location
- Chr. 13: 49912702 - 49936501 on Build GRCh38
- UniGene ID
- Hs.44235
- Species
- Homo sapiens
- Gene Symbol
- SVIL
- Entrez Gene ID
- 6840
- Gene Name
- supervillin
- Gene Aliases
- Chromosome Location
- Chr. 10: 29457338 - 29737001 on Build GRCh38
- UniGene ID
- Hs.499209
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK001792.1 |
- |
|
2026 |
|
- Gene Symbol
- TCF7
- Entrez Gene ID
- 6932
- Gene Name
- transcription factor 7 (T-cell specific, HMG-box)
- Gene Aliases
- TCF-1
- Chromosome Location
- Chr. 5: 134114703 - 134148229 on Build GRCh38
- UniGene ID
- Hs.573153
- Species
- Homo sapiens
- Gene Symbol
- TLCD2
- Entrez Gene ID
- 727910
- Gene Name
- TLC domain containing 2
- Gene Aliases
- Chromosome Location
- Chr. 17: 1702790 - 1710368 on Build GRCh38
- UniGene ID
- Hs.531005
- Species
- Homo sapiens
- Gene Symbol
- WDR27
- Entrez Gene ID
- 253769
- Gene Name
- WD repeat domain 27
- Gene Aliases
- Chromosome Location
- Chr. 6: 169426420 - 169702071 on Build GRCh38
- UniGene ID
- Hs.131903
- Species
- Homo sapiens
- Gene Symbol
- WT1-AS
- Entrez Gene ID
- 51352
- Gene Name
- WT1 antisense RNA
- Gene Aliases
- WIT-1, WIT1, WT1-AS1, WT1AS
- Chromosome Location
- Chr. 11: 32435518 - 32458769 on Build GRCh38
- UniGene ID
- Hs.567499
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX387533.2 |
- |
1 |
84 |
|
- Gene Symbol
- ZFY
- Entrez Gene ID
- 7544
- Gene Name
- zinc finger protein, Y-linked
- Gene Aliases
- ZNF911
- Chromosome Location
- Chr. Y: 2934402 - 2982508 on Build GRCh38
- UniGene ID
- Hs.522845
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK310553.1 |
- |
6 |
1620 |
|
- Gene Symbol
- ZMAT3
- Entrez Gene ID
- 64393
- Gene Name
- zinc finger matrin-type 3
- Gene Aliases
- PAG608, WIG-1, WIG1
- Chromosome Location
- Chr. 3: 179017223 - 179072279 on Build GRCh38
- UniGene ID
- Hs.371609
- Species
- Homo sapiens
- Gene Symbol
- ZNF556
- Entrez Gene ID
- 80032
- Gene Name
- zinc finger protein 556
- Gene Aliases
- Chromosome Location
- Chr. 19: 2867335 - 2878517 on Build GRCh38
- UniGene ID
- Hs.287433
- Species
- Homo sapiens
- Gene Symbol
- ZNF682
- Entrez Gene ID
- 91120
- Gene Name
- zinc finger protein 682
- Gene Aliases
- BC39498_3
- Chromosome Location
- Chr. 19: 20004418 - 20039468 on Build GRCh38
- UniGene ID
- Hs.306298
- Species
- Homo sapiens
- Gene Symbol
- ZNFX1
- Entrez Gene ID
- 57169
- Gene Name
- zinc finger NFX1-type containing 1
- Gene Aliases
- Chromosome Location
- Chr. 20: 49245897 - 49278244 on Build GRCh38
- UniGene ID
- Hs.371794
- Species
- Homo sapiens
View Full Product Details