Gene Transcripts
- Gene Symbol
- HDAC9
- Entrez Gene ID
- 9734
- Gene Name
- histone deacetylase 9
- Gene Aliases
- HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
- Chromosome Location
- Chr. 7: 18086942 - 19003745 on Build GRCh38
- UniGene ID
- Hs.196054
- Species
- Homo sapiens
- Gene Symbol
- AASDH
- Entrez Gene ID
- 132949
- Gene Name
- aminoadipate-semialdehyde dehydrogenase
- Gene Aliases
- ACSF4, LYS2, NRPS1098, NRPS998
- Chromosome Location
- Chr. 4: 56337687 - 56387677 on Build GRCh38
- UniGene ID
- Hs.104347
- Species
- Homo sapiens
- Gene Symbol
- ASB18
- Entrez Gene ID
- 401036
- Gene Name
- ankyrin repeat and SOCS box containing 18
- Gene Aliases
- ASB-18
- Chromosome Location
- Chr. 2: 236194872 - 236264345 on Build GRCh38
- UniGene ID
- Hs.639909
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123854.1 |
- |
5 |
2149 |
|
- Gene Symbol
- CLCC1
- Entrez Gene ID
- 23155
- Gene Name
- chloride channel CLIC like 1
- Gene Aliases
- MCLC
- Chromosome Location
- Chr. 1: 108929508 - 108963499 on Build GRCh38
- UniGene ID
- Hs.658489
- Species
- Homo sapiens
- Gene Symbol
- DENND6A
- Entrez Gene ID
- 201627
- Gene Name
- DENN domain containing 6A
- Gene Aliases
- AFI1A, FAM116A
- Chromosome Location
- Chr. 3: 57625454 - 57693118 on Build GRCh38
- UniGene ID
- Hs.91085
- Species
- Homo sapiens
- Gene Symbol
- FBXO40
- Entrez Gene ID
- 51725
- Gene Name
- F-box protein 40
- Gene Aliases
- FBX40
- Chromosome Location
- Chr. 3: 121593323 - 121630292 on Build GRCh38
- UniGene ID
- Hs.272564
- Species
- Homo sapiens
- Gene Symbol
- FLJ43879
- Entrez Gene ID
- 401039
- Gene Name
- FLJ43879 protein
- Gene Aliases
- Chromosome Location
- Chr. 2: 238919302 - 238926269 on Build GRCh38
- UniGene ID
- Hs.516791
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034162.1 |
|
4 |
2124 |
|
GenBank mRNA |
AK125867.1 |
- |
4 |
2124 |
|
- Gene Symbol
- GOSR1
- Entrez Gene ID
- 9527
- Gene Name
- golgi SNAP receptor complex member 1
- Gene Aliases
- GOLIM2, GOS-28, GOS28, GOS28/P28, GS28, P28
- Chromosome Location
- Chr. 17: 30477402 - 30528314 on Build GRCh38
- UniGene ID
- Hs.462680
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128816.1 |
- |
2 |
1755 |
|
- Gene Symbol
- HP09025
- Entrez Gene ID
- 100652929
- Gene Name
- uncharacterized LOC100652929
- Gene Aliases
- Chromosome Location
- Chr. 17: 79707266 - 79712317 on Build GRCh38
- UniGene ID
- Hs.559249
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_109783.1 |
|
2 |
2888 |
|
GenBank mRNA |
AB593148.1 |
- |
3 |
2786 |
|
- Gene Symbol
- JOSD1
- Entrez Gene ID
- 9929
- Gene Name
- Josephin domain containing 1
- Gene Aliases
- dJ508I15.2
- Chromosome Location
- Chr. 22: 38685543 - 38701418 on Build GRCh38
- UniGene ID
- Hs.3094
- Species
- Homo sapiens
- Gene Symbol
- KIAA1257
- Entrez Gene ID
- 57501
- Gene Name
- KIAA1257
- Gene Aliases
- Chromosome Location
- Chr. 3: 128910186 - 129032431 on Build GRCh38
- UniGene ID
- Hs.518247
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB033083.1 |
- |
8 |
5123 |
|
- Gene Symbol
- LINC00963
- Entrez Gene ID
- 100506190
- Gene Name
- long intergenic non-protein coding RNA 963
- Gene Aliases
- Chromosome Location
- Chr. 9: 129488660 - 129513686 on Build GRCh38
- UniGene ID
- Hs.529860
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057050.1 |
- |
1 |
1189 |
|
- Gene Symbol
- LYPD4
- Entrez Gene ID
- 147719
- Gene Name
- LY6/PLAUR domain containing 4
- Gene Aliases
- SMR
- Chromosome Location
- Chr. 19: 41833686 - 41844771 on Build GRCh38
- UniGene ID
- Hs.382361
- Species
- Homo sapiens
- Gene Symbol
- NAF1
- Entrez Gene ID
- 92345
- Gene Name
- nuclear assembly factor 1 ribonucleoprotein
- Gene Aliases
- Chromosome Location
- Chr. 4: 163109133 - 163170388 on Build GRCh38
- UniGene ID
- Hs.129095
- Species
- Homo sapiens
- Gene Symbol
- NFATC3
- Entrez Gene ID
- 4775
- Gene Name
- nuclear factor of activated T-cells 3
- Gene Aliases
- NFAT4, NFATX
- Chromosome Location
- Chr. 16: 68085366 - 68229259 on Build GRCh38
- UniGene ID
- Hs.436585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057574.1 |
- |
1 |
459 |
|
- Gene Symbol
- NFS1
- Entrez Gene ID
- 9054
- Gene Name
- NFS1 cysteine desulfurase
- Gene Aliases
- HUSSY-08, IscS, NIFS
- Chromosome Location
- Chr. 20: 35668688 - 35699365 on Build GRCh38
- UniGene ID
- Hs.194692
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK091049.1 |
- |
1 |
102 |
|
- Gene Symbol
- NUAK2
- Entrez Gene ID
- 81788
- Gene Name
- NUAK family kinase 2
- Gene Aliases
- SNARK
- Chromosome Location
- Chr. 1: 205302059 - 205321791 on Build GRCh38
- UniGene ID
- Hs.497512
- Species
- Homo sapiens
- Gene Symbol
- OSBPL2
- Entrez Gene ID
- 9885
- Gene Name
- oxysterol binding protein like 2
- Gene Aliases
- DFNA67, DNFA67, ORP-2, ORP2
- Chromosome Location
- Chr. 20: 62238485 - 62296213 on Build GRCh38
- UniGene ID
- Hs.473254
- Species
- Homo sapiens
- Gene Symbol
- PDK3
- Entrez Gene ID
- 5165
- Gene Name
- pyruvate dehydrogenase kinase 3
- Gene Aliases
- CMTX6, GS1-358P8.4
- Chromosome Location
- Chr. X: 24465227 - 24550466 on Build GRCh38
- UniGene ID
- Hs.296031
- Species
- Homo sapiens
- Gene Symbol
- PHACTR4
- Entrez Gene ID
- 65979
- Gene Name
- phosphatase and actin regulator 4
- Gene Aliases
- PPP1R124
- Chromosome Location
- Chr. 1: 28369574 - 28500369 on Build GRCh38
- UniGene ID
- Hs.225641
- Species
- Homo sapiens
- Gene Symbol
- RDH13
- Entrez Gene ID
- 112724
- Gene Name
- retinol dehydrogenase 13 (all-trans/9-cis)
- Gene Aliases
- SDR7C3
- Chromosome Location
- Chr. 19: 55038895 - 55069774 on Build GRCh38
- UniGene ID
- Hs.327631
- Species
- Homo sapiens
- Gene Symbol
- RIN1
- Entrez Gene ID
- 9610
- Gene Name
- Ras and Rab interactor 1
- Gene Aliases
- Chromosome Location
- Chr. 11: 66332063 - 66337235 on Build GRCh38
- UniGene ID
- Hs.1030
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209445.1 |
- |
7 |
4092 |
|
- Gene Symbol
- SLC22A5
- Entrez Gene ID
- 6584
- Gene Name
- solute carrier family 22 member 5
- Gene Aliases
- CDSP, OCTN2
- Chromosome Location
- Chr. 5: 132369704 - 132395614 on Build GRCh38
- UniGene ID
- Hs.443572
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209484.1 |
- |
2 |
3476 |
|
- Gene Symbol
- SMIM11A
- Entrez Gene ID
- 54065
- Gene Name
- small integral membrane protein 11A
- Gene Aliases
- C21orf51, FAM165B, SMIM11
- Chromosome Location
- Chr. 21: 34375450 - 34402777 on Build GRCh38
- UniGene ID
- Hs.656195
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BM989806.1 |
- |
1 |
307 |
|
- Gene Symbol
- SNX15
- Entrez Gene ID
- 29907
- Gene Name
- sorting nexin 15
- Gene Aliases
- HSAF001435
- Chromosome Location
- Chr. 11: 65027408 - 65040572 on Build GRCh38
- UniGene ID
- Hs.80132
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209439.1 |
- |
6 |
1594 |
|
- Gene Symbol
- STIL
- Entrez Gene ID
- 6491
- Gene Name
- SCL/TAL1 interrupting locus
- Gene Aliases
- MCPH7, SIL
- Chromosome Location
- Chr. 1: 47250139 - 47314787 on Build GRCh38
- UniGene ID
- Hs.525198
- Species
- Homo sapiens
- Gene Symbol
- TMEM170B
- Entrez Gene ID
- 100113407
- Gene Name
- transmembrane protein 170B
- Gene Aliases
- Chromosome Location
- Chr. 6: 11538042 - 11583524 on Build GRCh38
- UniGene ID
- Hs.146317
- Species
- Homo sapiens
- Gene Symbol
- TSPAN5
- Entrez Gene ID
- 10098
- Gene Name
- tetraspanin 5
- Gene Aliases
- NET-4, NET4, TM4SF9, TSPAN-5
- Chromosome Location
- Chr. 4: 98470367 - 98658661 on Build GRCh38
- UniGene ID
- Hs.118118
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126702.1 |
- |
1 |
617 |
|
- Gene Symbol
- ZNF17
- Entrez Gene ID
- 7565
- Gene Name
- zinc finger protein 17
- Gene Aliases
- HPF3, KOX10
- Chromosome Location
- Chr. 19: 57409252 - 57421939 on Build GRCh38
- UniGene ID
- Hs.729403
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB075827.1 |
- |
1 |
2889 |
|
- Gene Symbol
- ZNF578
- Entrez Gene ID
- 147660
- Gene Name
- zinc finger protein 578
- Gene Aliases
- Chromosome Location
- Chr. 19: 52453514 - 52516878 on Build GRCh38
- UniGene ID
- Hs.157287
- Species
- Homo sapiens
- Gene Symbol
- ZNF681
- Entrez Gene ID
- 148213
- Gene Name
- zinc finger protein 681
- Gene Aliases
- Chromosome Location
- Chr. 19: 23739195 - 23758891 on Build GRCh38
- UniGene ID
- Hs.399952
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138286.2 |
NP_612143.2
|
4 |
3401 |
|
- Gene Symbol
- ZNF789
- Entrez Gene ID
- 285989
- Gene Name
- zinc finger protein 789
- Gene Aliases
- Chromosome Location
- Chr. 7: 99472819 - 99487600 on Build GRCh38
- UniGene ID
- Hs.440384
- Species
- Homo sapiens
- Gene Symbol
- ZSCAN29
- Entrez Gene ID
- 146050
- Gene Name
- zinc finger and SCAN domain containing 29
- Gene Aliases
- ZNF690, Zfp690
- Chromosome Location
- Chr. 15: 43358172 - 43371032 on Build GRCh38
- UniGene ID
- Hs.418287
- Species
- Homo sapiens
View Full Product Details