Gene Transcripts
- Gene Symbol
- ZNF737
- Entrez Gene ID
- 100129842
- Gene Name
- zinc finger protein 737
- Gene Aliases
- ZNF102
- Chromosome Location
- Chr. 19: 20531230 - 20565821 on Build GRCh38
- UniGene ID
- Hs.515696
- Species
- Homo sapiens
- Gene Symbol
- LOC105372319
- Entrez Gene ID
- 105372319
- Gene Name
- putative zinc finger protein 56
- Gene Aliases
- Chromosome Location
- Chr. 19: 20662592 - 20692266 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC400682
- Entrez Gene ID
- 400682
- Gene Name
- zinc finger protein 100-like
- Gene Aliases
- Chromosome Location
- Chr. 19: 21637371 - 21658948 on Build GRCh38
- UniGene ID
- Hs.568075
- Species
- Homo sapiens
- Gene Symbol
- LOC441666
- Entrez Gene ID
- 441666
- Gene Name
- zinc finger protein 91 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 10: 42331866 - 42368045 on Build GRCh38
- UniGene ID
- Hs.255729
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024380.1 |
|
3 |
1667 |
|
GenBank mRNA |
BX537675.1 |
- |
1 |
201 |
|
- Gene Symbol
- LOC732272
- Entrez Gene ID
- 732272
- Gene Name
- uncharacterized LOC732272
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.369632
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092688.1 |
- |
5 |
1112 |
|
- Gene Symbol
- ZNF100
- Entrez Gene ID
- 163227
- Gene Name
- zinc finger protein 100
- Gene Aliases
- Chromosome Location
- Chr. 19: 21724041 - 21767628 on Build GRCh38
- UniGene ID
- Hs.365142
- Species
- Homo sapiens
- Gene Symbol
- ZNF107
- Entrez Gene ID
- 51427
- Gene Name
- zinc finger protein 107
- Gene Aliases
- Y8, ZFD25, ZNF588, smap-7
- Chromosome Location
- Chr. 7: 64665012 - 64711582 on Build GRCh38
- UniGene ID
- Hs.50216
- Species
- Homo sapiens
- Gene Symbol
- ZNF117
- Entrez Gene ID
- 51351
- Gene Name
- zinc finger protein 117
- Gene Aliases
- H-plk, HPF9
- Chromosome Location
- Chr. 7: 64974452 - 64991036 on Build GRCh38
- UniGene ID
- Hs.250693
- Species
- Homo sapiens
- Gene Symbol
- ZNF138
- Entrez Gene ID
- 7697
- Gene Name
- zinc finger protein 138
- Gene Aliases
- pHZ-32
- Chromosome Location
- Chr. 7: 64794388 - 64853800 on Build GRCh38
- UniGene ID
- Hs.184080
- Species
- Homo sapiens
- Gene Symbol
- ZNF141
- Entrez Gene ID
- 7700
- Gene Name
- zinc finger protein 141
- Gene Aliases
- D4S90, PAPA6, pHZ-44
- Chromosome Location
- Chr. 4: 336995 - 377071 on Build GRCh38
- UniGene ID
- Hs.654355
- Species
- Homo sapiens
- Gene Symbol
- ZNF208
- Entrez Gene ID
- 7757
- Gene Name
- zinc finger protein 208
- Gene Aliases
- PMIDP, ZNF95
- Chromosome Location
- Chr. 19: 21966095 - 22010990 on Build GRCh38
- UniGene ID
- Hs.732975
- Species
- Homo sapiens
- Gene Symbol
- ZNF253
- Entrez Gene ID
- 56242
- Gene Name
- zinc finger protein 253
- Gene Aliases
- BMZF-1, BMZF1, ZNF411
- Chromosome Location
- Chr. 19: 19865905 - 19893484 on Build GRCh38
- UniGene ID
- Hs.301059
- Species
- Homo sapiens
- Gene Symbol
- ZNF254
- Entrez Gene ID
- 9534
- Gene Name
- zinc finger protein 254
- Gene Aliases
- BMZF-5, HD-ZNF1, ZNF539, ZNF91L
- Chromosome Location
- Chr. 19: 24033405 - 24129968 on Build GRCh38
- UniGene ID
- Hs.434406
- Species
- Homo sapiens
- Gene Symbol
- ZNF257
- Entrez Gene ID
- 113835
- Gene Name
- zinc finger protein 257
- Gene Aliases
- BMZF-4, BMZF4
- Chromosome Location
- Chr. 19: 22052448 - 22091103 on Build GRCh38
- UniGene ID
- Hs.283900
- Species
- Homo sapiens
- Gene Symbol
- ZNF273
- Entrez Gene ID
- 10793
- Gene Name
- zinc finger protein 273
- Gene Aliases
- HZF9
- Chromosome Location
- Chr. 7: 64882493 - 64937316 on Build GRCh38
- UniGene ID
- Hs.520889
- Species
- Homo sapiens
- Gene Symbol
- ZNF429
- Entrez Gene ID
- 353088
- Gene Name
- zinc finger protein 429
- Gene Aliases
- Chromosome Location
- Chr. 19: 21505257 - 21541157 on Build GRCh38
- UniGene ID
- Hs.572567
- Species
- Homo sapiens
- Gene Symbol
- ZNF43
- Entrez Gene ID
- 7594
- Gene Name
- zinc finger protein 43
- Gene Aliases
- HTF6, KOX27, ZNF39L1
- Chromosome Location
- Chr. 19: 21804946 - 21852095 on Build GRCh38
- UniGene ID
- Hs.534365
- Species
- Homo sapiens
- Gene Symbol
- ZNF430
- Entrez Gene ID
- 80264
- Gene Name
- zinc finger protein 430
- Gene Aliases
- Chromosome Location
- Chr. 19: 21020620 - 21060046 on Build GRCh38
- UniGene ID
- Hs.729202
- Species
- Homo sapiens
- Gene Symbol
- ZNF431
- Entrez Gene ID
- 170959
- Gene Name
- zinc finger protein 431
- Gene Aliases
- Chromosome Location
- Chr. 19: 21142009 - 21191367 on Build GRCh38
- UniGene ID
- Hs.656394
- Species
- Homo sapiens
- Gene Symbol
- ZNF479
- Entrez Gene ID
- 90827
- Gene Name
- zinc finger protein 479
- Gene Aliases
- HKr19, KR19
- Chromosome Location
- Chr. 7: 57119574 - 57139899 on Build GRCh38
- UniGene ID
- Hs.616660
- Species
- Homo sapiens
- Gene Symbol
- ZNF486
- Entrez Gene ID
- 90649
- Gene Name
- zinc finger protein 486
- Gene Aliases
- KRBO2
- Chromosome Location
- Chr. 19: 20167214 - 20200490 on Build GRCh38
- UniGene ID
- Hs.590991
- Species
- Homo sapiens
- Gene Symbol
- ZNF492
- Entrez Gene ID
- 57615
- Gene Name
- zinc finger protein 492
- Gene Aliases
- ZNF115
- Chromosome Location
- Chr. 19: 22634324 - 22667670 on Build GRCh38
- UniGene ID
- Hs.232108
- Species
- Homo sapiens
- Gene Symbol
- ZNF493
- Entrez Gene ID
- 284443
- Gene Name
- zinc finger protein 493
- Gene Aliases
- Chromosome Location
- Chr. 19: 21397119 - 21427495 on Build GRCh38
- UniGene ID
- Hs.656558
- Species
- Homo sapiens
- Gene Symbol
- ZNF506
- Entrez Gene ID
- 440515
- Gene Name
- zinc finger protein 506
- Gene Aliases
- Chromosome Location
- Chr. 19: 19792711 - 19821751 on Build GRCh38
- UniGene ID
- Hs.351906
- Species
- Homo sapiens
- Gene Symbol
- ZNF595
- Entrez Gene ID
- 152687
- Gene Name
- zinc finger protein 595
- Gene Aliases
- Chromosome Location
- Chr. 4: 53276 - 88211 on Build GRCh38
- UniGene ID
- Hs.709469
- Species
- Homo sapiens
- Gene Symbol
- ZNF626
- Entrez Gene ID
- 199777
- Gene Name
- zinc finger protein 626
- Gene Aliases
- Chromosome Location
- Chr. 19: 20619939 - 20661596 on Build GRCh38
- UniGene ID
- Hs.657001
- Species
- Homo sapiens
- Gene Symbol
- ZNF675
- Entrez Gene ID
- 171392
- Gene Name
- zinc finger protein 675
- Gene Aliases
- TBZF, TIZ
- Chromosome Location
- Chr. 19: 23651079 - 23687220 on Build GRCh38
- UniGene ID
- Hs.264345
- Species
- Homo sapiens
- Gene Symbol
- ZNF676
- Entrez Gene ID
- 163223
- Gene Name
- zinc finger protein 676
- Gene Aliases
- Chromosome Location
- Chr. 19: 22179101 - 22196951 on Build GRCh38
- UniGene ID
- Hs.55452
- Species
- Homo sapiens
- Gene Symbol
- ZNF678
- Entrez Gene ID
- 339500
- Gene Name
- zinc finger protein 678
- Gene Aliases
- Chromosome Location
- Chr. 1: 227563519 - 227677443 on Build GRCh38
- UniGene ID
- Hs.30323
- Species
- Homo sapiens
- Gene Symbol
- ZNF679
- Entrez Gene ID
- 168417
- Gene Name
- zinc finger protein 679
- Gene Aliases
- Chromosome Location
- Chr. 7: 64228474 - 64266931 on Build GRCh38
- UniGene ID
- Hs.520884
- Species
- Homo sapiens
- Gene Symbol
- ZNF680
- Entrez Gene ID
- 340252
- Gene Name
- zinc finger protein 680
- Gene Aliases
- Chromosome Location
- Chr. 7: 64500253 - 64563135 on Build GRCh38
- UniGene ID
- Hs.520886
- Species
- Homo sapiens
- Gene Symbol
- ZNF681
- Entrez Gene ID
- 148213
- Gene Name
- zinc finger protein 681
- Gene Aliases
- Chromosome Location
- Chr. 19: 23739195 - 23758891 on Build GRCh38
- UniGene ID
- Hs.399952
- Species
- Homo sapiens
- Gene Symbol
- ZNF682
- Entrez Gene ID
- 91120
- Gene Name
- zinc finger protein 682
- Gene Aliases
- BC39498_3
- Chromosome Location
- Chr. 19: 20004418 - 20039468 on Build GRCh38
- UniGene ID
- Hs.306298
- Species
- Homo sapiens
- Gene Symbol
- ZNF695
- Entrez Gene ID
- 57116
- Gene Name
- zinc finger protein 695
- Gene Aliases
- SBZF3
- Chromosome Location
- Chr. 1: 246945547 - 247008093 on Build GRCh38
- UniGene ID
- Hs.669893
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020394.4 |
NP_065127.4
|
4 |
1580 |
|
- Gene Symbol
- ZNF708
- Entrez Gene ID
- 7562
- Gene Name
- zinc finger protein 708
- Gene Aliases
- KOX8, ZNF15, ZNF15L1
- Chromosome Location
- Chr. 19: 21290388 - 21329439 on Build GRCh38
- UniGene ID
- Hs.466296
- Species
- Homo sapiens
- Gene Symbol
- ZNF714
- Entrez Gene ID
- 148206
- Gene Name
- zinc finger protein 714
- Gene Aliases
- Chromosome Location
- Chr. 19: 21082147 - 21125094 on Build GRCh38
- UniGene ID
- Hs.729186
- Species
- Homo sapiens
- Gene Symbol
- ZNF716
- Entrez Gene ID
- 441234
- Gene Name
- zinc finger protein 716
- Gene Aliases
- Chromosome Location
- Chr. 7: 57450177 - 57473559 on Build GRCh38
- UniGene ID
- Hs.533121
- Species
- Homo sapiens
- Gene Symbol
- ZNF718
- Entrez Gene ID
- 255403
- Gene Name
- zinc finger protein 718
- Gene Aliases
- Chromosome Location
- Chr. 4: 124247 - 202306 on Build GRCh38
- UniGene ID
- Hs.636638
- Species
- Homo sapiens
- Gene Symbol
- ZNF724P
- Entrez Gene ID
- 440519
- Gene Name
- zinc finger protein 724, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 19: 23223805 - 23250370 on Build GRCh38
- UniGene ID
- Hs.675814
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK301230.1 |
- |
4 |
1387 |
|
- Gene Symbol
- ZNF726
- Entrez Gene ID
- 730087
- Gene Name
- zinc finger protein 726
- Gene Aliases
- Chromosome Location
- Chr. 19: 23914882 - 23933967 on Build GRCh38
- UniGene ID
- Hs.655305
- Species
- Homo sapiens
- Gene Symbol
- ZNF727
- Entrez Gene ID
- 442319
- Gene Name
- zinc finger protein 727
- Gene Aliases
- ZNF727P
- Chromosome Location
- Chr. 7: 64044806 - 64085184 on Build GRCh38
- UniGene ID
- Hs.640774
- Species
- Homo sapiens
- Gene Symbol
- ZNF728
- Entrez Gene ID
- 388523
- Gene Name
- zinc finger protein 728
- Gene Aliases
- Chromosome Location
- Chr. 19: 22974883 - 23003208 on Build GRCh38
- UniGene ID
- Hs.616766
- Species
- Homo sapiens
- Gene Symbol
- ZNF729
- Entrez Gene ID
- 100287226
- Gene Name
- zinc finger protein 729
- Gene Aliases
- Chromosome Location
- Chr. 19: 22286404 - 22317438 on Build GRCh38
- UniGene ID
- Hs.531629
- Species
- Homo sapiens
- Gene Symbol
- ZNF730
- Entrez Gene ID
- 100129543
- Gene Name
- zinc finger protein 730
- Gene Aliases
- Chromosome Location
- Chr. 19: 23074990 - 23147886 on Build GRCh38
- UniGene ID
- Hs.427120
- Species
- Homo sapiens
- Gene Symbol
- ZNF732
- Entrez Gene ID
- 654254
- Gene Name
- zinc finger protein 732
- Gene Aliases
- Chromosome Location
- Chr. 4: 270675 - 305321 on Build GRCh38
- UniGene ID
- Hs.698668
- Species
- Homo sapiens
- Gene Symbol
- ZNF733P
- Entrez Gene ID
- 643955
- Gene Name
- zinc finger protein 733, pseudogene
- Gene Aliases
- ZNF733
- Chromosome Location
- Chr. 7: 63291287 - 63304056 on Build GRCh38
- UniGene ID
- Hs.583308
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003952.1 |
|
3 |
1452 |
|
- Gene Symbol
- ZNF736
- Entrez Gene ID
- 728927
- Gene Name
- zinc finger protein 736
- Gene Aliases
- Chromosome Location
- Chr. 7: 64313873 - 64356634 on Build GRCh38
- UniGene ID
- Hs.386162
- Species
- Homo sapiens
- Gene Symbol
- ZNF738
- Entrez Gene ID
- 148203
- Gene Name
- zinc finger protein 738
- Gene Aliases
- Chromosome Location
- Chr. 19: 21358933 - 21388582 on Build GRCh38
- UniGene ID
- Hs.124384
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027130.1 |
|
5 |
7054 |
|
- Gene Symbol
- ZNF85
- Entrez Gene ID
- 7639
- Gene Name
- zinc finger protein 85
- Gene Aliases
- HPF4, HTF1
- Chromosome Location
- Chr. 19: 20923227 - 20950697 on Build GRCh38
- UniGene ID
- Hs.37138
- Species
- Homo sapiens
- Gene Symbol
- ZNF876P
- Entrez Gene ID
- 642280
- Gene Name
- zinc finger protein 876, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 4: 212600 - 255985 on Build GRCh38
- UniGene ID
- Hs.673877
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027481.1 |
|
2 |
808 |
|
GenBank mRNA |
BC028359.1 |
- |
2 |
803 |
|
BC046475.1 |
- |
2 |
778 |
|
- Gene Symbol
- ZNF90
- Entrez Gene ID
- 7643
- Gene Name
- zinc finger protein 90
- Gene Aliases
- HTF9
- Chromosome Location
- Chr. 19: 20077994 - 20127032 on Build GRCh38
- UniGene ID
- Hs.55304
- Species
- Homo sapiens
- Gene Symbol
- ZNF91
- Entrez Gene ID
- 7644
- Gene Name
- zinc finger protein 91
- Gene Aliases
- HPF7, HTF10
- Chromosome Location
- Chr. 19: 23305004 - 23395560 on Build GRCh38
- UniGene ID
- Hs.531632
- Species
- Homo sapiens
- Gene Symbol
- ZNF92
- Entrez Gene ID
- 168374
- Gene Name
- zinc finger protein 92
- Gene Aliases
- HEL-203, HPF12, HTF12, TF12
- Chromosome Location
- Chr. 7: 65373799 - 65401135 on Build GRCh38
- UniGene ID
- Hs.9521
- Species
- Homo sapiens
- Gene Symbol
- ZNF93
- Entrez Gene ID
- 81931
- Gene Name
- zinc finger protein 93
- Gene Aliases
- HPF34, HTF34, TF34, ZNF505
- Chromosome Location
- Chr. 19: 19900913 - 19935573 on Build GRCh38
- UniGene ID
- Hs.301059
- Species
- Homo sapiens
- Gene Symbol
- ZNF98
- Entrez Gene ID
- 148198
- Gene Name
- zinc finger protein 98
- Gene Aliases
- F7175, ZNF739
- Chromosome Location
- Chr. 19: 22391011 - 22422628 on Build GRCh38
- UniGene ID
- Hs.667355
- Species
- Homo sapiens
- Gene Symbol
- ZNF99
- Entrez Gene ID
- 7652
- Gene Name
- zinc finger protein 99
- Gene Aliases
- C19orf9, F8281
- Chromosome Location
- Chr. 19: 22752183 - 22784171 on Build GRCh38
- UniGene ID
- Hs.568380
- Species
- Homo sapiens
View Full Product Details