Gene Transcripts
- Gene Symbol
- HIST2H4A
- Entrez Gene ID
- 8370
- Gene Name
- histone cluster 2, H4a
- Gene Aliases
- FO108, H4, H4/n, H4F2, H4FN, HIST2H4
- Chromosome Location
- Chr. 1: 149832657 - 149833052 on Build GRCh38
- UniGene ID
- Hs.55468
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC019846.1 |
- |
2 |
684 |
|
BC108260.1 |
- |
2 |
1442 |
|
View Full Product Details