Gene Transcripts

Gene Symbol
TP53TG3B
Entrez Gene ID
729355
Gene Name
TP53 target 3B
Gene Aliases
TP53TG3E, TP53TG3F
Chromosome Location
Chr. 16: 33358385 - 33363478 on Build GRCh38
UniGene ID
Hs.592038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099687.3 NP_001093157.1 1 130
NR_110897.1 1 130
NR_110911.1 1 130
XM_006721084.1 XP_006721147.1 1 104
XM_011545933.1 XP_011544235.1 1 104
XM_017023638.1 XP_016879127.1 1 2622
XM_017023639.1 XP_016879128.1 1 111

Gene Symbol
LOC102723655
Entrez Gene ID
102723655
Gene Name
TP53-target gene 3 protein
Gene Aliases
Chromosome Location
Chr. 16|NT_187383: 14735 - 19833 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006724904.3 XP_006724967.1 1 2627
XM_006724905.3 XP_006724968.1 1 2623
XM_017030123.1 XP_016885612.1 1 2622
XM_017030124.1 XP_016885613.1 1 2622

Gene Symbol
LOC102723713
Entrez Gene ID
102723713
Gene Name
TP53-target gene 3 protein
Gene Aliases
Chromosome Location
Chr. 16|NT_187383: 933861 - 936780 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011546237.1 XP_011544539.1 1 442
XM_011546238.1 XP_011544540.1 1 442
XM_017030126.1 XP_016885615.1 1 442
XM_017030127.1 XP_016885616.1 1 442

Gene Symbol
TP53TG3
Entrez Gene ID
24150
Gene Name
TP53 target 3
Gene Aliases
P53TG3, TP53TG3A, TP53TG3E, TP53TG3F
Chromosome Location
Chr. 16: 32673528 - 32678621 on Build GRCh38
UniGene ID
Hs.592038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016212.4 NP_057296.1 1 130
NR_110886.1 1 130
NR_110910.1 1 130
XM_006721026.1 XP_006721089.1 1 104
XM_011545779.1 XP_011544081.1 1 104
XM_017023102.1 XP_016878591.1 1 2622
XM_017023103.1 XP_016878592.1 1 111
GenBank mRNA AB023508.1 - 1 130
AK312220.1 - 1 111
AL133588.1 - 1 733
BC096733.1 - 1 128 40015196
BC098165.1 - 1 128 40015195
BX381390.2 - 1 23

Gene Symbol
TP53TG3C
Entrez Gene ID
653550
Gene Name
TP53 target 3C
Gene Aliases
TP53TG3, TP53TG3E, TP53TG3F
Chromosome Location
Chr. 16: 33191770 - 33196858 on Build GRCh38
UniGene ID
Hs.592038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205259.2 NP_001192188.1 1 128
NR_110914.1 1 128
XM_006721074.2 XP_006721137.1 1 104
XM_011545920.1 XP_011544222.1 1 104
XM_017023591.1 XP_016879080.1 1 2622
XM_017023592.1 XP_016879081.1 1 111
XM_017023593.1 XP_016879082.1 1 111
GenBank mRNA DB053188.1 - 1 12

Gene Symbol
TP53TG3D
Entrez Gene ID
729264
Gene Name
TP53 target 3D
Gene Aliases
TP53TG3, TP53TG3B, TP53TG3C, TP53TG3E, TP53TG3F
Chromosome Location
Chr. 16: 32251926 - 32255928 on Build GRCh38
UniGene ID
Hs.513537
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243722.2 NP_001230651.1 1 128
NR_110898.1 1 128
XM_017023633.1 XP_016879122.1 1 1525
XM_017023634.1 XP_016879123.1 1 1525
XM_017023635.1 XP_016879124.1 1 1526
XM_017023636.1 XP_016879125.1 1 1526
XM_017023637.1 XP_016879126.1 1 1526
GenBank mRNA AB023509.1 - 1 129
BC096700.1 - 1 128 40015193
BC099842.1 - 1 128 40015192
CR541650.1 - 1 98
DA077802.1 - 1 108

Gene Symbol
TP53TG3E
Entrez Gene ID
102724101
Gene Name
TP53 target 3 family member E
Gene Aliases
Chromosome Location
Chr. 16: 33301849 - 33306938 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006721117.2 XP_006721180.1 1 104
XM_011546032.1 XP_011544334.1 1 104
XM_017023950.1 XP_016879439.1 1 2622
XM_017023951.1 XP_016879440.1 1 2622
XM_017023952.1 XP_016879441.1 1 111
XM_017023953.1 XP_016879442.1 1 111
XM_017023954.1 XP_016879443.1 1 2623

Gene Symbol
TP53TG3F
Entrez Gene ID
102724127
Gene Name
TP53 target 3 family member F
Gene Aliases
Chromosome Location
Chr. 16: 33457155 - 33462249 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006721118.1 XP_006721181.1 1 104
XM_011546033.1 XP_011544335.1 1 104
XM_017023955.1 XP_016879444.1 1 2622
XM_017023956.1 XP_016879445.1 1 2622
XM_017023957.1 XP_016879446.1 1 111
XM_017023958.1 XP_016879447.1 1 111
XM_017023959.1 XP_016879448.1 1 2623

View Full Product Details