Gene Transcripts
- Gene Symbol
- LOC441666
- Entrez Gene ID
- 441666
- Gene Name
- zinc finger protein 91 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 10: 42331866 - 42368045 on Build GRCh38
- UniGene ID
- Hs.255729
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024380.1 |
|
3 |
2071 |
|
GenBank mRNA |
BX537675.1 |
- |
1 |
605 |
|
View Full Product Details