Gene Transcripts
- Gene Symbol
- PAQR7
- Entrez Gene ID
- 164091
- Gene Name
- progestin and adipoQ receptor family member 7
- Gene Aliases
- MPRA, PGLP, mSR
- Chromosome Location
- Chr. 1: 25861484 - 25876707 on Build GRCh38
- UniGene ID
- Hs.523652
- Species
- Homo sapiens
- Gene Symbol
- AAK1
- Entrez Gene ID
- 22848
- Gene Name
- AP2 associated kinase 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 69457995 - 69643845 on Build GRCh38
- UniGene ID
- Hs.468878
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB028971.1 |
- |
18 |
3319 |
|
- Gene Symbol
- CARS
- Entrez Gene ID
- 833
- Gene Name
- cysteinyl-tRNA synthetase
- Gene Aliases
- CARS1, CYSRS, MGC:11246
- Chromosome Location
- Chr. 11: 3000922 - 3057451 on Build GRCh38
- UniGene ID
- Hs.274873
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB208814.1 |
- |
12 |
4541 |
|
- Gene Symbol
- CLINT1
- Entrez Gene ID
- 9685
- Gene Name
- clathrin interactor 1
- Gene Aliases
- CLINT, ENTH, EPN4, EPNR
- Chromosome Location
- Chr. 5: 157785743 - 157859175 on Build GRCh38
- UniGene ID
- Hs.644000
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC030264.1 |
- |
1 |
224 |
|
- Gene Symbol
- DEF8
- Entrez Gene ID
- 54849
- Gene Name
- differentially expressed in FDCP 8 homolog (mouse)
- Gene Aliases
- Chromosome Location
- Chr. 16: 89948697 - 89968060 on Build GRCh38
- UniGene ID
- Hs.62771
- Species
- Homo sapiens
- Gene Symbol
- EIF3G
- Entrez Gene ID
- 8666
- Gene Name
- eukaryotic translation initiation factor 3 subunit G
- Gene Aliases
- EIF3-P42, EIF3S4, eIF3-delta, eIF3-p44
- Chromosome Location
- Chr. 19: 10115014 - 10119923 on Build GRCh38
- UniGene ID
- Hs.529059
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK309944.1 |
- |
5 |
551 |
|
AK315896.1 |
- |
5 |
557 |
|
- Gene Symbol
- GIT2
- Entrez Gene ID
- 9815
- Gene Name
- GIT ArfGAP 2
- Gene Aliases
- CAT-2, CAT2, PKL
- Chromosome Location
- Chr. 12: 109929792 - 109996389 on Build GRCh38
- UniGene ID
- Hs.434996
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125441.1 |
- |
1 |
353 |
|
- Gene Symbol
- GOLGA4
- Entrez Gene ID
- 2803
- Gene Name
- golgin A4
- Gene Aliases
- CRPF46, GCP2, GOLG, MU-RMS-40.18, p230
- Chromosome Location
- Chr. 3: 37243191 - 37366879 on Build GRCh38
- UniGene ID
- Hs.344151
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX640663.1 |
- |
2 |
4003 |
|
- Gene Symbol
- LOC101927727
- Entrez Gene ID
- 101927727
- Gene Name
- uncharacterized LOC101927727
- Gene Aliases
- Chromosome Location
- Chr. 17: 909632 - 911212 on Build GRCh38
- UniGene ID
- Hs.666198
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135633.1 |
|
1 |
34 |
|
GenBank mRNA |
BQ934941.1 |
- |
1 |
34 |
|
- Gene Symbol
- LOC102724738
- Entrez Gene ID
- 102724738
- Gene Name
- solute carrier family 25 member 1 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 7: 63346800 - 63350066 on Build GRCh38
- UniGene ID
- Hs.512544
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_136256.1 |
|
1 |
1736 |
|
GenBank mRNA |
BC036215.1 |
- |
1 |
1476 |
|
- Gene Symbol
- LOC728323
- Entrez Gene ID
- 728323
- Gene Name
- uncharacterized LOC728323
- Gene Aliases
- Chromosome Location
- Chr. 2: 242088633 - 242160503 on Build GRCh38
- UniGene ID
- Hs.380737
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125248.1 |
- |
7 |
1300 |
|
- Gene Symbol
- MCOLN2
- Entrez Gene ID
- 255231
- Gene Name
- mucolipin 2
- Gene Aliases
- TRP-ML2, TRPML2
- Chromosome Location
- Chr. 1: 84925583 - 84997122 on Build GRCh38
- UniGene ID
- Hs.591446
- Species
- Homo sapiens
- Gene Symbol
- PPWD1
- Entrez Gene ID
- 23398
- Gene Name
- peptidylprolyl isomerase domain and WD repeat containing 1
- Gene Aliases
- Chromosome Location
- Chr. 5: 65563236 - 65587549 on Build GRCh38
- UniGene ID
- Hs.121432
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093675.1 |
- |
1 |
1153 |
|
- Gene Symbol
- SMU1
- Entrez Gene ID
- 55234
- Gene Name
- DNA replication regulator and spliceosomal factor
- Gene Aliases
- BWD, SMU-1, fSAP57
- Chromosome Location
- Chr. 9: 33041852 - 33076716 on Build GRCh38
- UniGene ID
- Hs.655351
- Species
- Homo sapiens
- Gene Symbol
- SNHG17
- Entrez Gene ID
- 388796
- Gene Name
- small nucleolar RNA host gene 17
- Gene Aliases
- Chromosome Location
- Chr. 20: 38420592 - 38435375 on Build GRCh38
- UniGene ID
- Hs.400876
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX640887.1 |
- |
4 |
3831 |
|
- Gene Symbol
- TACC1
- Entrez Gene ID
- 6867
- Gene Name
- transforming acidic coiled-coil containing protein 1
- Gene Aliases
- Ga55
- Chromosome Location
- Chr. 8: 38728186 - 38853028 on Build GRCh38
- UniGene ID
- Hs.279245
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124297.1 |
- |
3 |
307 |
|
- Gene Symbol
- TOR4A
- Entrez Gene ID
- 54863
- Gene Name
- torsin family 4 member A
- Gene Aliases
- C9orf167
- Chromosome Location
- Chr. 9: 137277828 - 137282641 on Build GRCh38
- UniGene ID
- Hs.495541
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK000392.1 |
- |
2 |
1103 |
|
- Gene Symbol
- ZNF335
- Entrez Gene ID
- 63925
- Gene Name
- zinc finger protein 335
- Gene Aliases
- MCPH10, NIF-1, NIF1, NIF2
- Chromosome Location
- Chr. 20: 45948653 - 45972910 on Build GRCh38
- UniGene ID
- Hs.174193
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054694.1 |
- |
2 |
1626 |
|
- Gene Symbol
- ZNF839
- Entrez Gene ID
- 55778
- Gene Name
- zinc finger protein 839
- Gene Aliases
- C14orf131
- Chromosome Location
- Chr. 14: 102317377 - 102343174 on Build GRCh38
- UniGene ID
- Hs.743359
- Species
- Homo sapiens
View Full Product Details