Gene Transcripts
- Gene Symbol
- LOC100131257
- Entrez Gene ID
- 100131257
- Gene Name
- zinc finger protein 655 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 7: 7075770 - 7096786 on Build GRCh38
- UniGene ID
- Hs.551110
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034022.1 |
|
1 |
9723 |
|
- Gene Symbol
- ALOX5
- Entrez Gene ID
- 240
- Gene Name
- arachidonate 5-lipoxygenase
- Gene Aliases
- 5-LO, 5-LOX, 5LPG, LOG5
- Chromosome Location
- Chr. 10: 45374166 - 45446121 on Build GRCh38
- UniGene ID
- Hs.89499
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB208946.1 |
- |
3 |
1016 |
|
- Gene Symbol
- LINC01609
- Entrez Gene ID
- 101927487
- Gene Name
- long intergenic non-protein coding RNA 1609
- Gene Aliases
- Chromosome Location
- Chr. 8: 111098961 - 111236203 on Build GRCh38
- UniGene ID
- Hs.630004
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BI459905.1 |
- |
2 |
394 |
|
- Gene Symbol
- LOC441666
- Entrez Gene ID
- 441666
- Gene Name
- zinc finger protein 91 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 10: 42331866 - 42368045 on Build GRCh38
- UniGene ID
- Hs.255729
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK131313.1 |
- |
6 |
1992 |
|
- Gene Symbol
- MIPOL1
- Entrez Gene ID
- 145282
- Gene Name
- mirror-image polydactyly 1
- Gene Aliases
- CCDC193
- Chromosome Location
- Chr. 14: 37197888 - 37579207 on Build GRCh38
- UniGene ID
- Hs.660396
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX647795.1 |
- |
10 |
3629 |
|
- Gene Symbol
- SMC5-AS1
- Entrez Gene ID
- 100507299
- Gene Name
- SMC5 antisense RNA 1 (head to head)
- Gene Aliases
- MAMDC2-AS2
- Chromosome Location
- Chr. 9: 70216059 - 70258874 on Build GRCh38
- UniGene ID
- Hs.556816
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CR936772.1 |
- |
3 |
3826 |
|
View Full Product Details