Gene Transcripts

Gene Symbol
Rfk
Entrez Gene ID
499328
Gene Name
riboflavin kinase
Gene Aliases
Chromosome Location
Chr. 1: 236494221 - 236501733 on Build Rnor_6.0
UniGene ID
Rn.13360
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014106.1 NP_001014128.1 2 430
GenBank mRNA BC079125.1 - 2 430
FQ219206.1 - 2 398
FQ223869.1 - 2 443

View Full Product Details