Gene Transcripts
- Gene Symbol
- PDCD6IPP2
- Entrez Gene ID
- 646278
- Gene Name
- PDCD6IP pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 15: 28789834 - 28856574 on Build GRCh38
- UniGene ID
- Hs.590143
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037599.1 |
|
9 |
1115 |
|
GenBank mRNA |
DB073810.1 |
- |
3 |
289 |
|
View Full Product Details