Gene Transcripts

Gene Symbol
Usp11
Entrez Gene ID
408217
Gene Name
ubiquitin specific peptidase 11
Gene Aliases
Uhx1
Chromosome Location
Chr. X: 1687820 - 1703991 on Build Rnor_6.0
UniGene ID
Rn.2492
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008861.2 NP_001008861.2 12 1505
GenBank mRNA BC090333.1 - 12 1505
BC105613.1 - 8 918

View Full Product Details