Gene Transcripts
- Gene Symbol
- NBPF13P
- Entrez Gene ID
- 644861
- Gene Name
- neuroblastoma breakpoint family member 13, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 1: 147099482 - 147114346 on Build GRCh38
- UniGene ID
- Hs.451804
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103466.1 |
|
5, 6 |
731 |
|
GenBank mRNA |
BX336604.2 |
- |
5, 6 |
731 |
|
CB154168.1 |
- |
3, 4 |
408 |
|
View Full Product Details