Gene Transcripts

Gene Symbol
Lrrc8c
Entrez Gene ID
289443
Gene Name
leucine rich repeat containing 8 family, member C
Gene Aliases
RGD1306585
Chromosome Location
Chr. 14: 5283655 - 5378698 on Build Rnor_6.0
UniGene ID
Rn.25285
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037179.1 NP_001032256.1 4 1904
XM_006250542.2 XP_006250604.1 2 1637
GenBank mRNA BC100076.1 - 4 1904

View Full Product Details