Gene Transcripts

Gene Symbol
LINC01101
Entrez Gene ID
84931
Gene Name
long intergenic non-protein coding RNA 1101
Gene Aliases
Chromosome Location
Chr. 2: 120464335 - 120466349 on Build GRCh38
UniGene ID
Hs.334519
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027181.1 1 561
GenBank mRNA AK027722.1 - 1 333
BC112205.1 - 1 441
BC113708.1 - 1 429

View Full Product Details