Gene Transcripts

Gene Symbol
KIF1A
Entrez Gene ID
547
Gene Name
kinesin family member 1A
Gene Aliases
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
Chromosome Location
Chr. 2: 240713764 - 240821025 on Build GRCh38
UniGene ID
Hs.516802
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244008.1 NP_001230937.1 49 8846
NM_001320705.1 NP_001307634.1 48 8570
NM_004321.6 NP_004312.2 46 8356
XM_005247022.1 XP_005247079.1 50 8784
XM_005247024.1 XP_005247081.1 49 8757
XM_005247027.1 XP_005247084.1 48 8478
XM_005247028.2 XP_005247085.1 47 8457
XM_006712605.1 XP_006712668.1 48 8730
XM_011511364.1 XP_011509666.1 50 8796
XM_011511365.1 XP_011509667.1 49 8508
XM_011511366.2 XP_011509668.1 40 7856
XM_011511367.1 XP_011509669.1 40 7786
XM_017004383.1 XP_016859872.1 50 8859
XM_017004384.1 XP_016859873.1 50 8871
XM_017004385.1 XP_016859874.1 50 8856
XM_017004386.1 XP_016859875.1 49 8832
XM_017004387.1 XP_016859876.1 49 8583
XM_017004388.1 XP_016859877.1 49 8580
XM_017004389.1 XP_016859878.1 48 8556
XM_017004390.1 XP_016859879.1 48 8481
XM_017004391.1 XP_016859880.1 49 8635
GenBank mRNA AB209138.1 - 22 5655
AB210029.1 - 44 5236
AB290172.1 - 49 8846
AK054791.1 - 1 1491
BC009491.2 - 1 1578
BC021199.2 - 1 1201
BX537556.1 - 9 5159

View Full Product Details