Gene Transcripts

Gene Symbol
ZNF660
Entrez Gene ID
285349
Gene Name
zinc finger protein 660
Gene Aliases
Chromosome Location
Chr. 3: 44584847 - 44599699 on Build GRCh38
UniGene ID
Hs.733134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173658.2 NP_775929.2 3 405
XM_005265090.4 XP_005265147.1 2 503
XM_006713117.3 XP_006713180.1 3 504
XM_011533622.2 XP_011531924.1 3 708
GenBank mRNA AK094189.1 - 3 405
BC075094.2 - 1 96 30915605
BC075095.2 - 1 96 30915393

View Full Product Details