Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001265587.1 | NP_001252516.1 | 3 | 524 | |
NM_005543.3 | NP_005534.2 | 2 | 429 | ||
GenBank mRNA | AK302780.1 | - | 2 | 371 | |
AY082014.1 | - | 3 | 524 | ||
BC032810.1 | - | 2 | 359 | ||
BC040435.1 | - | 2 | 359 | ||
BC053345.1 | - | 2 | 359 | ||
BC061886.1 | - | 2 | 382 | ||
BC071706.1 | - | 2 | 362 | 5167223 | |
BC106721.1 | - | 2 | 361 | 40014433 | |
BC106722.1 | - | 2 | 366 | 40014435 | |
BC107045.2 | - | 1 | 55 | ||
BX334591.2 | - | 2 | 372 | ||
S72482.1 | - | 2 | 368 |