Gene Transcripts
- Gene Symbol
- CLRN1
- Entrez Gene ID
- 7401
- Gene Name
- clarin 1
- Gene Aliases
- RP61, USH3, USH3A
- Chromosome Location
- Chr. 3: 150926163 - 150972999 on Build GRCh38
- UniGene ID
- Hs.745448
- Species
- Homo sapiens
- Gene Symbol
- FAM188B2
- Entrez Gene ID
- 646951
- Gene Name
- family with sequence similarity 188 member B2
- Gene Aliases
- C3orf76, CLRN1
- Chromosome Location
- Chr. 3: 150871030 - 150891522 on Build GRCh38
- UniGene ID
- Hs.380222
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
HM626137.1 |
- |
|
11 |
|
HM626138.1 |
- |
|
11 |
|
View Full Product Details