Gene Transcripts

Gene Symbol
D130043K22Rik
Entrez Gene ID
210108
Gene Name
RIKEN cDNA D130043K22 gene
Gene Aliases
4930451E12Rik, Kiaa0319
Chromosome Location
Chr. 13: 24843930 - 24902159 on Build GRCm38
UniGene ID
Mm.261514
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK051381.1 - 3 396
AK122246.1 - 3 393
BC115723.1 - 3 312 40086618
BC115940.1 - 3 312 40086616
BY718582.1 - 3 400

View Full Product Details