Gene Transcripts

Gene Symbol
4921524L21Rik
Entrez Gene ID
70901
Gene Name
RIKEN cDNA 4921524L21 gene
Gene Aliases
Chromosome Location
Chr. 18: 6603633 - 6640435 on Build GRCm38
UniGene ID
Mm.158484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027598.1 NP_081874.1 10 1243
GenBank mRNA AK014961.1 - 10 1244
AK161257.1 - 625
BC049627.1 - 8 1088 6706381

View Full Product Details