Gene Transcripts

Gene Symbol
4930579G24Rik
Entrez Gene ID
75939
Gene Name
RIKEN cDNA 4930579G24 gene
Gene Aliases
Chromosome Location
Chr. 3: 79629079 - 79632820 on Build GRCm38
UniGene ID
Mm.221520
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029482.1 NP_083758.1 2 572
GenBank mRNA AK016321.1 - 2 543
AK137776.1 - 562
BC052483.1 - 2 553

View Full Product Details