Gene Transcripts

Gene Symbol
Idh3a
Entrez Gene ID
114096
Gene Name
isocitrate dehydrogenase 3 (NAD+) alpha
Gene Aliases
Chromosome Location
Chr. 8: 59164601 - 59183899 on Build Rnor_6.0
UniGene ID
Rn.95104
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053638.1 NP_446090.1 4 281
GenBank mRNA AB047541.1 - 4 281

View Full Product Details