Gene Transcripts

Gene Symbol
Dscam
Entrez Gene ID
171119
Gene Name
Down syndrome cell adhesion molecule
Gene Aliases
Chromosome Location
Chr. 11: 37004776 - 37599866 on Build Rnor_6.0
UniGene ID
Rn.24399
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133587.1 NP_598271.1 2 676
GenBank mRNA AF334385.1 - 2 676

View Full Product Details