Gene Transcripts

Gene Symbol
Scaf8
Entrez Gene ID
245926
Gene Name
SR-related CTD-associated factor 8
Gene Aliases
Rbm16
Chromosome Location
Chr. 1: 44018966 - 44120619 on Build Rnor_6.0
UniGene ID
Rn.10529
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139094.1 NP_620794.1 10, 11 1443
XM_006227858.2 XP_006227920.1 11, 12 1370
GenBank mRNA U49055.1 - 10, 11 1443

View Full Product Details