Gene Transcripts

Gene Symbol
Cdx4
Entrez Gene ID
12592
Gene Name
caudal type homeobox 4
Gene Aliases
Cdx-3, Cdx-4, Cdx3
Chromosome Location
Chr. X: 103321398 - 103330224 on Build GRCm38
UniGene ID
Mm.4353
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007674.3 NP_031700.1 3 845
GenBank mRNA AK141681.1 - 3 259
BC119158.1 - 3 833 8733907
L08061.1 - 3 848

View Full Product Details