Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114339.2 | NP_001107811.1 | 2 | 1020 | |
NM_023792.2 | NP_076281.1 | 2 | 320 | ||
GenBank mRNA | AF200357.1 | - | 2 | 320 | |
AF347700.1 | - | 2 | 1018 | ||
AK017345.1 | - | 2 | 887 | ||
AK050222.1 | - | 2 | 321 | ||
BC023496.1 | - | 2 | 242 | 5054239 |