Gene Transcripts
- Gene Symbol
- Rtca
- Entrez Gene ID
- 66368
- Gene Name
- RNA 3'-terminal phosphate cyclase
- Gene Aliases
- 2310009A18Rik, AI450277, RPC, Rtcd1
- Chromosome Location
- Chr. 3: 116488963 - 116508175 on Build GRCm38
- UniGene ID
- Mm.251228
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025517.3 |
NP_079793.2
|
2, 3 |
340 |
|
GenBank mRNA |
AK002543.1 |
- |
2, 3 |
251 |
|
AK009245.1 |
- |
2, 3 |
276 |
|
AK151108.1 |
- |
2, 3 |
228 |
|
AK151400.1 |
- |
2, 3 |
228 |
|
AK152129.1 |
- |
2, 3 |
341 |
|
AK159961.1 |
- |
2, 3 |
230 |
|
AK167830.1 |
- |
2, 3 |
217 |
|
BC016519.1 |
- |
2, 3 |
266 |
3495638 |
BC090403.1 |
- |
2, 3 |
311 |
5293337 |
View Full Product Details