Gene Transcripts

Gene Symbol
Olfr698
Entrez Gene ID
258595
Gene Name
olfactory receptor 698
Gene Aliases
MOR283-3
Chromosome Location
Chr. 7: 106752451 - 106753386 on Build GRCm38
UniGene ID
Mm.377585
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146602.2 NP_666813.2 1 248
GenBank mRNA BC113955.1 - 1 247
BC148217.1 - 1 244

View Full Product Details