Gene Transcripts

Gene Symbol
Olfr473
Entrez Gene ID
258771
Gene Name
olfactory receptor 473
Gene Aliases
MOR204-4
Chromosome Location
Chr. 7: 107933522 - 107934454 on Build GRCm38
UniGene ID
Mm.451662
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146775.1 NP_666986.1 1 824
GenBank mRNA BC148218.1 - 1 828

View Full Product Details