Gene Transcripts

Gene Symbol
Fbxl17
Entrez Gene ID
50758
Gene Name
F-box and leucine-rich repeat protein 17
Gene Aliases
6330576B01Rik, AI452053, BB073797, C130023C01Rik, Fbx13, Fbxo13
Chromosome Location
Chr. 17: 63045949 - 63504042 on Build GRCm38
UniGene ID
Mm.136366
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015794.1 NP_056609.1 9 14570
GenBank mRNA AK047946.1 - 1 771
AK086201.1 - 1 804
AK086431.1 - 1 578

View Full Product Details