Gene Transcripts

Gene Symbol
TRIM5
Entrez Gene ID
85363
Gene Name
tripartite motif containing 5
Gene Aliases
RNF88, TRIM5alpha
Chromosome Location
Chr. 11: 5590906 - 5685109 on Build GRCh38
UniGene ID
Hs.125300
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB362928.1 - 1, 2 407
AF220027.1 - 1, 2 469
AF220028.1 - 1, 2 469
AK027593.1 - 2, 3 664
DQ288685.1 - 1, 2 407

View Full Product Details