Gene Transcripts

Gene Symbol
LINC00698
Entrez Gene ID
285401
Gene Name
long intergenic non-protein coding RNA 698
Gene Aliases
Chromosome Location
Chr. 3: 63102688 - 63125062 on Build GRCh38
UniGene ID
Hs.211125
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027104.1 7 1556
GenBank mRNA BC039502.1 - 6 1148
BC043407.1 - 9 1677
BC104474.1 - 1 458
BC104475.1 - 1 458

View Full Product Details