Gene Transcripts
- Gene Symbol
- BMX
- Entrez Gene ID
- 660
- Gene Name
- BMX non-receptor tyrosine kinase
- Gene Aliases
- ETK, PSCTK2, PSCTK3
- Chromosome Location
- Chr. X: 15500777 - 15556529 on Build GRCh38
- UniGene ID
- Hs.495731
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320866.1 |
NP_001307795.1
|
3, 4 |
417 |
|
NM_001721.6 |
NP_001712.1
|
3, 4 |
355 |
|
NM_203281.2 |
NP_975010.1
|
3, 4 |
417 |
|
XM_017029752.1 |
XP_016885241.1
|
3, 4 |
387 |
|
GenBank mRNA |
AF045459.1 |
- |
3, 4 |
431 |
|
AK291824.1 |
- |
3, 4 |
387 |
|
AK313568.1 |
- |
3, 4 |
350 |
|
BC016652.2 |
- |
3, 4 |
372 |
3919253 |
BP236451.1 |
- |
3, 4 |
417 |
|
BQ226991.1 |
- |
3, 4 |
377 |
|
CB998198.1 |
- |
|
392 |
|
X83107.1 |
- |
3, 4 |
277 |
|
View Full Product Details