Gene Transcripts
- Gene Symbol
- TUFT1
- Entrez Gene ID
- 7286
- Gene Name
- tuftelin 1
- Gene Aliases
- Chromosome Location
- Chr. 1: 151540294 - 151583583 on Build GRCh38
- UniGene ID
- Hs.489922
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001126337.1 |
NP_001119809.1
|
10, 11 |
987 |
|
NM_001301317.1 |
NP_001288246.1
|
12, 13 |
1197 |
|
NM_020127.2 |
NP_064512.1
|
11, 12 |
1062 |
|
XM_011509961.2 |
XP_011508263.1
|
9, 10 |
896 |
|
XM_017002223.1 |
XP_016857712.1
|
11, 12 |
1138 |
|
XM_017002224.1 |
XP_016857713.1
|
10, 11 |
1259 |
|
GenBank mRNA |
AF254260.1 |
- |
11, 12 |
1062 |
|
AK026989.1 |
- |
4, 5 |
316 |
|
AK075001.1 |
- |
10, 11 |
984 |
|
AK223161.1 |
- |
11, 12 |
1058 |
|
AK298066.1 |
- |
12, 13 |
1197 |
|
AK299925.1 |
- |
10, 11 |
975 |
|
AK315414.1 |
- |
11, 12 |
1058 |
|
AL136917.1 |
- |
11, 12 |
1050 |
|
BC002933.1 |
- |
10, 11 |
955 |
3956873 |
BC008301.1 |
- |
11, 12 |
1042 |
3506004 |
View Full Product Details