Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 612885-613379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF7
Entrez Gene ID
3665
Gene Name
interferon regulatory factor 7
Gene Aliases
IMD39, IRF-7H, IRF7A, IRF7B, IRF7C, IRF7H
UniGene
Hs.166120
Gene Chromosome Location
Chr.11: 612555-615999 on Build GRCh38
Transcripts
NM_001572.3 NM_004029.2 NM_004031.2 XM_005252906.3 XM_005252907.3 XM_005252909.3 XM_011520066.2 XM_017017674.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1254936 c.1128G>A p.G376G Chr.11: 613354-613354 on Build GRCh38
123781 c.1318C>T p.R440C Chr.11: 613076-613076 on Build GRCh38
4461609 c.1213C>G p.P405A Chr.11: 613269-613269 on Build GRCh38
4461610 c.1174C>G p.P392A Chr.11: 613269-613269 on Build GRCh38
6048411 c.1064delC p.P355fs*97 Chr.11: 613379-613379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889002024 Chr.11: 612922-612922 on Build GRCh38
rs571178819 Chr.11: 612909-612909 on Build GRCh38
rs572167003 Chr.11: 612949-612949 on Build GRCh38
rs971448511 Chr.11: 613352-613380 on Build GRCh38
rs970393690 Chr.11: 613353-613354 on Build GRCh38
rs773112663 Chr.11: 613042-613042 on Build GRCh38
rs749678457 Chr.11: 613299-613299 on Build GRCh38
rs757013967 Chr.11: 612953-612953 on Build GRCh38
rs375323253 Chr.11: 613094-613094 on Build GRCh38
rs767736050 Chr.11: 613289-613289 on Build GRCh38
rs771545271 Chr.11: 613259-613259 on Build GRCh38
rs766875876 Chr.11: 612991-612991 on Build GRCh38
rs1131665 Chr.11: 613208-613208 on Build GRCh38
rs786205223 Chr.11: 613215-613215 on Build GRCh38
rs777558624 Chr.11: 612951-612951 on Build GRCh38
rs759300160 Chr.11: 613333-613333 on Build GRCh38
rs772688443 Chr.11: 612978-612978 on Build GRCh38
rs773863600 Chr.11: 613181-613181 on Build GRCh38
rs1021187471 Chr.11: 613294-613294 on Build GRCh38
rs368825184 Chr.11: 612992-612992 on Build GRCh38
rs1061505 Chr.11: 613297-613297 on Build GRCh38
rs368953784 Chr.11: 613302-613302 on Build GRCh38
rs746520294 Chr.11: 613321-613321 on Build GRCh38
rs760536724 Chr.11: 613278-613278 on Build GRCh38
rs773984280 Chr.11: 613252-613252 on Build GRCh38
rs373729529 Chr.11: 613018-613018 on Build GRCh38
rs755335495 Chr.11: 613300-613300 on Build GRCh38
rs944667389 Chr.11: 613103-613103 on Build GRCh38
rs993328518 Chr.11: 613171-613171 on Build GRCh38
rs748115929 Chr.11: 612971-612971 on Build GRCh38
rs757506612 Chr.11: 613287-613287 on Build GRCh38
rs767789503 Chr.11: 613348-613348 on Build GRCh38
rs373431859 Chr.11: 612994-612994 on Build GRCh38
rs780994139 Chr.11: 613052-613052 on Build GRCh38
rs750448993 Chr.11: 613290-613290 on Build GRCh38
rs916189739 Chr.11: 612945-612945 on Build GRCh38
rs201165872 Chr.11: 613076-613076 on Build GRCh38
rs747554292 Chr.11: 613083-613083 on Build GRCh38
rs549942229 Chr.11: 613270-613270 on Build GRCh38
rs746276782 Chr.11: 613143-613143 on Build GRCh38
rs772011524 Chr.11: 613075-613075 on Build GRCh38
rs762797158 Chr.11: 613053-613053 on Build GRCh38
rs763997590 Chr.11: 613066-613070 on Build GRCh38
rs535754355 Chr.11: 613213-613213 on Build GRCh38
rs148420621 Chr.11: 613023-613023 on Build GRCh38
rs779918847 Chr.11: 613343-613344 on Build GRCh38
rs759270303 Chr.11: 613008-613008 on Build GRCh38
rs751540818 Chr.11: 613161-613162 on Build GRCh38
rs757383689 Chr.11: 613227-613227 on Build GRCh38
rs747031092 Chr.11: 613130-613130 on Build GRCh38
rs371536958 Chr.11: 612964-612964 on Build GRCh38
rs781777277 Chr.11: 613228-613228 on Build GRCh38
rs749608310 Chr.11: 613241-613241 on Build GRCh38
rs749532271 Chr.11: 612958-612958 on Build GRCh38
rs1051390 Chr.11: 613165-613165 on Build GRCh38
rs771501527 Chr.11: 613086-613086 on Build GRCh38
rs576205574 Chr.11: 612919-612919 on Build GRCh38
rs936370311 Chr.11: 612928-612928 on Build GRCh38
rs764365065 Chr.11: 613219-613219 on Build GRCh38
rs992330485 Chr.11: 612998-612998 on Build GRCh38
rs900460497 Chr.11: 613249-613249 on Build GRCh38
rs138601170 Chr.11: 613115-613115 on Build GRCh38
rs748323177 Chr.11: 613071-613071 on Build GRCh38
rs1039452743 Chr.11: 613238-613238 on Build GRCh38
rs751755858 Chr.11: 613222-613222 on Build GRCh38
rs772725374 Chr.11: 613318-613318 on Build GRCh38
rs761174337 Chr.11: 612989-612989 on Build GRCh38
rs756964153 Chr.11: 613176-613195 on Build GRCh38
rs369313404 Chr.11: 612915-612915 on Build GRCh38
rs747818359 Chr.11: 613307-613307 on Build GRCh38
rs535695919 Chr.11: 612924-612924 on Build GRCh38
rs200063173 Chr.11: 612999-612999 on Build GRCh38
rs774975476 Chr.11: 613092-613092 on Build GRCh38
rs371452982 Chr.11: 613235-613235 on Build GRCh38
rs11246213 Chr.11: 612967-612967 on Build GRCh38
rs767504897 Chr.11: 613185-613185 on Build GRCh38
rs979593621 Chr.11: 613309-613309 on Build GRCh38
rs373690626 Chr.11: 613111-613111 on Build GRCh38
rs145109888 Chr.11: 613324-613324 on Build GRCh38
rs768248464 Chr.11: 613250-613250 on Build GRCh38
rs143070036 Chr.11: 613097-613097 on Build GRCh38
rs760896372 Chr.11: 613164-613164 on Build GRCh38
rs548071462 Chr.11: 613168-613168 on Build GRCh38
rs993689673 Chr.11: 612937-612937 on Build GRCh38
rs764450622 Chr.11: 613285-613285 on Build GRCh38
rs746890167 Chr.11: 612952-612952 on Build GRCh38
rs189985591 Chr.11: 613144-613144 on Build GRCh38
rs142523418 Chr.11: 613102-613102 on Build GRCh38
rs942117850 Chr.11: 613237-613237 on Build GRCh38
rs758532865 Chr.11: 612973-612973 on Build GRCh38
rs752888043 Chr.11: 613124-613124 on Build GRCh38
rs764133862 Chr.11: 613022-613022 on Build GRCh38
rs555431855 Chr.11: 612927-612927 on Build GRCh38
rs760800493 Chr.11: 613358-613358 on Build GRCh38
rs945551950 Chr.11: 613047-613047 on Build GRCh38
rs957792868 Chr.11: 613332-613332 on Build GRCh38
rs577986906 Chr.11: 612972-612972 on Build GRCh38
rs368955367 Chr.11: 613112-613112 on Build GRCh38
rs149039709 Chr.11: 613087-613087 on Build GRCh38
rs760975524 Chr.11: 612944-612944 on Build GRCh38
rs747693922 Chr.11: 613257-613257 on Build GRCh38
rs765147452 Chr.11: 613337-613337 on Build GRCh38
rs948990982 Chr.11: 613158-613158 on Build GRCh38
rs556610785 Chr.11: 612918-612918 on Build GRCh38
rs775126859 Chr.11: 613091-613091 on Build GRCh38
rs897657995 Chr.11: 612936-612936 on Build GRCh38
rs12422022 Chr.11: 613192-613192 on Build GRCh38
rs376213301 Chr.11: 613196-613196 on Build GRCh38
rs755896373 Chr.11: 613114-613114 on Build GRCh38
rs771624398 Chr.11: 613131-613131 on Build GRCh38
rs762128931 Chr.11: 613340-613340 on Build GRCh38
rs781763496 Chr.11: 613136-613136 on Build GRCh38
rs150387655 Chr.11: 613017-613017 on Build GRCh38
rs141451948 Chr.11: 613005-613005 on Build GRCh38
rs761295533 Chr.11: 613183-613183 on Build GRCh38
rs375116104 Chr.11: 613020-613020 on Build GRCh38
rs746399119 Chr.11: 613231-613231 on Build GRCh38
rs555493535 Chr.11: 613282-613282 on Build GRCh38
rs780303221 Chr.11: 612957-612957 on Build GRCh38
rs1030581700 Chr.11: 613070-613070 on Build GRCh38
rs771649294 Chr.11: 613315-613315 on Build GRCh38
rs377416331 Chr.11: 613096-613096 on Build GRCh38
rs745452747 Chr.11: 613256-613259 on Build GRCh38
rs778988142 Chr.11: 613067-613067 on Build GRCh38
rs781061158 Chr.11: 613218-613219 on Build GRCh38
rs144501906 Chr.11: 612986-612986 on Build GRCh38
rs776600480 Chr.11: 613325-613325 on Build GRCh38
rs750526448 Chr.11: 613354-613354 on Build GRCh38
rs758608826 Chr.11: 613127-613127 on Build GRCh38
rs767381892 Chr.11: 613110-613110 on Build GRCh38
rs753604903 Chr.11: 613206-613206 on Build GRCh38
rs755203402 Chr.11: 613057-613057 on Build GRCh38
rs779495519 Chr.11: 613338-613338 on Build GRCh38
rs755795955 Chr.11: 613291-613292 on Build GRCh38
rs572572807 Chr.11: 613284-613284 on Build GRCh38

View Full Product Details