Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31575480-31575974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNF
Entrez Gene ID
7124
Gene Name
tumor necrosis factor
Gene Aliases
DIF, TNF-alpha, TNFA, TNFSF2, TNLG1F
UniGene
Hs.241570
Gene Chromosome Location
Chr.6: 31575567-31578336 on Build GRCh38
Transcripts
NM_000594.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3624579 c.21C>T p.I7I Chr.6: 31575762-31575762 on Build GRCh38
4734841 c.130G>A p.A44T Chr.6: 31575871-31575871 on Build GRCh38
3374203 c.99C>T p.L33L Chr.6: 31575840-31575840 on Build GRCh38
5560328 c.3G>A p.M1I Chr.6: 31575744-31575744 on Build GRCh38
4610793 c.40G>A p.E14K Chr.6: 31575781-31575781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144480538 Chr.6: 31575879-31575879 on Build GRCh38
rs904527578 Chr.6: 31575636-31575636 on Build GRCh38
rs771650207 Chr.6: 31575755-31575755 on Build GRCh38
rs777874746 Chr.6: 31575910-31575910 on Build GRCh38
rs1054920809 Chr.6: 31575565-31575565 on Build GRCh38
rs541347219 Chr.6: 31575882-31575882 on Build GRCh38
rs201328097 Chr.6: 31575627-31575628 on Build GRCh38
rs867435711 Chr.6: 31575871-31575871 on Build GRCh38
rs4647199 Chr.6: 31575605-31575606 on Build GRCh38
rs953747858 Chr.6: 31575751-31575751 on Build GRCh38
rs55978323 Chr.6: 31575546-31575546 on Build GRCh38
rs369163834 Chr.6: 31575870-31575870 on Build GRCh38
rs779966486 Chr.6: 31575726-31575726 on Build GRCh38
rs948780599 Chr.6: 31575875-31575875 on Build GRCh38
rs377338702 Chr.6: 31575797-31575797 on Build GRCh38
rs750426362 Chr.6: 31575715-31575715 on Build GRCh38
rs982610810 Chr.6: 31575766-31575766 on Build GRCh38
rs757794184 Chr.6: 31575853-31575853 on Build GRCh38
rs775506502 Chr.6: 31575781-31575781 on Build GRCh38
rs916435207 Chr.6: 31575729-31575729 on Build GRCh38
rs554060079 Chr.6: 31575710-31575710 on Build GRCh38
rs756182468 Chr.6: 31575902-31575902 on Build GRCh38
rs2228088 Chr.6: 31575828-31575828 on Build GRCh38
rs1049309348 Chr.6: 31575867-31575868 on Build GRCh38
rs999084577 Chr.6: 31575673-31575673 on Build GRCh38
rs4645838 Chr.6: 31575634-31575635 on Build GRCh38
rs374531985 Chr.6: 31575763-31575763 on Build GRCh38
rs146375573 Chr.6: 31575789-31575789 on Build GRCh38
rs915225183 Chr.6: 31575842-31575842 on Build GRCh38
rs539666421 Chr.6: 31575523-31575523 on Build GRCh38
rs961336693 Chr.6: 31575548-31575548 on Build GRCh38
rs375864945 Chr.6: 31575736-31575736 on Build GRCh38
rs747406665 Chr.6: 31575838-31575838 on Build GRCh38
rs3179060 Chr.6: 31575895-31575895 on Build GRCh38
rs372719825 Chr.6: 31575712-31575712 on Build GRCh38
rs576621666 Chr.6: 31575827-31575827 on Build GRCh38
rs939173504 Chr.6: 31575550-31575550 on Build GRCh38
rs377627218 Chr.6: 31575768-31575768 on Build GRCh38
rs779870893 Chr.6: 31575825-31575825 on Build GRCh38
rs764955335 Chr.6: 31575814-31575814 on Build GRCh38
rs753713562 Chr.6: 31575805-31575805 on Build GRCh38
rs2515924 Chr.6: 31575714-31575714 on Build GRCh38
rs765178016 Chr.6: 31575926-31575926 on Build GRCh38
rs747694851 Chr.6: 31575754-31575754 on Build GRCh38
rs565862537 Chr.6: 31575756-31575756 on Build GRCh38
rs201502336 Chr.6: 31575764-31575764 on Build GRCh38
rs754639425 Chr.6: 31575811-31575811 on Build GRCh38
rs369780852 Chr.6: 31575743-31575743 on Build GRCh38
rs998337111 Chr.6: 31575735-31575735 on Build GRCh38
rs141667614 Chr.6: 31575861-31575861 on Build GRCh38
rs1012980674 Chr.6: 31575628-31575628 on Build GRCh38
rs759378982 Chr.6: 31575924-31575924 on Build GRCh38
rs755847921 Chr.6: 31575718-31575718 on Build GRCh38
rs538552423 Chr.6: 31575682-31575682 on Build GRCh38
rs368044930 Chr.6: 31575934-31575934 on Build GRCh38
rs558678940 Chr.6: 31575862-31575862 on Build GRCh38
rs766034823 Chr.6: 31575804-31575804 on Build GRCh38
rs3093660 Chr.6: 31575629-31575629 on Build GRCh38
rs770023902 Chr.6: 31575771-31575771 on Build GRCh38
rs759696973 Chr.6: 31575769-31575769 on Build GRCh38
rs1036935762 Chr.6: 31575880-31575880 on Build GRCh38
rs752785007 Chr.6: 31575709-31575709 on Build GRCh38
rs61761335 Chr.6: 31575597-31575597 on Build GRCh38
rs4248164 Chr.6: 31575571-31575571 on Build GRCh38
rs765073823 Chr.6: 31575513-31575513 on Build GRCh38
rs972459518 Chr.6: 31575784-31575784 on Build GRCh38
rs372568141 Chr.6: 31575699-31575699 on Build GRCh38
rs927783186 Chr.6: 31575535-31575535 on Build GRCh38
rs777405140 Chr.6: 31575836-31575836 on Build GRCh38
rs776753689 Chr.6: 31575912-31575912 on Build GRCh38
rs537401710 Chr.6: 31575514-31575514 on Build GRCh38
rs200241887 Chr.6: 31575878-31575878 on Build GRCh38
rs796223691 Chr.6: 31575633-31575634 on Build GRCh38

View Full Product Details