Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31576385-31576656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNF
Entrez Gene ID
7124
Gene Name
tumor necrosis factor
Gene Aliases
DIF, TNF-alpha, TNFA, TNFSF2, TNLG1F
UniGene
Hs.241570
Gene Chromosome Location
Chr.6: 31575567-31578336 on Build GRCh38
Transcripts
NM_000594.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7000215 c.195G>T p.R65S Chr.6: 31576542-31576542 on Build GRCh38
4885958 c.217C>G p.L73V Chr.6: 31576564-31576564 on Build GRCh38
3777486 c.231C>T p.V77V Chr.6: 31576578-31576578 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768947455 Chr.6: 31576523-31576523 on Build GRCh38
rs3093663 Chr.6: 31576448-31576448 on Build GRCh38
rs1010734656 Chr.6: 31576441-31576441 on Build GRCh38
rs577329144 Chr.6: 31576455-31576455 on Build GRCh38
rs541437699 Chr.6: 31576459-31576459 on Build GRCh38
rs139617507 Chr.6: 31576443-31576443 on Build GRCh38
rs775365890 Chr.6: 31576579-31576579 on Build GRCh38
rs759609722 Chr.6: 31576552-31576552 on Build GRCh38
rs762461783 Chr.6: 31576585-31576585 on Build GRCh38
rs972350853 Chr.6: 31576567-31576567 on Build GRCh38
rs906170945 Chr.6: 31576413-31576413 on Build GRCh38
rs769643681 Chr.6: 31576572-31576572 on Build GRCh38
rs896882312 Chr.6: 31576419-31576419 on Build GRCh38
rs1031304022 Chr.6: 31576425-31576425 on Build GRCh38
rs569914957 Chr.6: 31576446-31576446 on Build GRCh38
rs947775487 Chr.6: 31576411-31576411 on Build GRCh38
rs1038557595 Chr.6: 31576414-31576414 on Build GRCh38
rs536414350 Chr.6: 31576440-31576440 on Build GRCh38
rs780604770 Chr.6: 31576609-31576610 on Build GRCh38
rs746433040 Chr.6: 31576504-31576504 on Build GRCh38
rs35131721 Chr.6: 31576538-31576538 on Build GRCh38
rs763838774 Chr.6: 31576590-31576590 on Build GRCh38
rs772331441 Chr.6: 31576539-31576539 on Build GRCh38
rs780403381 Chr.6: 31576507-31576507 on Build GRCh38
rs751100224 Chr.6: 31576492-31576492 on Build GRCh38
rs373391738 Chr.6: 31576605-31576605 on Build GRCh38
rs559043897 Chr.6: 31576454-31576454 on Build GRCh38
rs1023506640 Chr.6: 31576463-31576463 on Build GRCh38
rs3093662 Chr.6: 31576412-31576412 on Build GRCh38
rs967964242 Chr.6: 31576521-31576521 on Build GRCh38
rs756466639 Chr.6: 31576506-31576506 on Build GRCh38
rs761373748 Chr.6: 31576624-31576624 on Build GRCh38
rs757664880 Chr.6: 31576500-31576500 on Build GRCh38
rs993943883 Chr.6: 31576437-31576437 on Build GRCh38
rs892302854 Chr.6: 31576436-31576436 on Build GRCh38
rs776692566 Chr.6: 31576543-31576543 on Build GRCh38
rs751210763 Chr.6: 31576620-31576620 on Build GRCh38
rs56340803 Chr.6: 31576632-31576632 on Build GRCh38
rs774257243 Chr.6: 31576530-31576530 on Build GRCh38
rs768258025 Chr.6: 31576619-31576619 on Build GRCh38
rs956294956 Chr.6: 31576565-31576565 on Build GRCh38
rs749414660 Chr.6: 31576522-31576522 on Build GRCh38
rs186267383 Chr.6: 31576631-31576631 on Build GRCh38
rs781640653 Chr.6: 31576503-31576503 on Build GRCh38

View Full Product Details