Gene Transcripts

Gene Symbol
Gas8
Entrez Gene ID
104346
Gene Name
growth arrest specific 8
Gene Aliases
Gas11
Chromosome Location
Chr. 8: 123518835 - 123536657 on Build GRCm38
UniGene ID
Mm.117265
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018855.2 NP_061343.2 11 1427
GenBank mRNA AY692443.1 - 11 1426
BC003779.1 - 11 1342 3592695
U19859.1 - 11 1448

View Full Product Details