Gene Transcripts

Gene Symbol
Palld
Entrez Gene ID
72333
Gene Name
palladin, cytoskeletal associated protein
Gene Aliases
2410003B16Rik, 6030492A02
Chromosome Location
Chr. 8: 61511429 - 61902700 on Build GRCm38
UniGene ID
Mm.29933
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081390.1 NP_001074859.1 2 776
NM_001293772.1 NP_001280701.1 2 780
GenBank mRNA AK052157.1 - 779
AK052489.1 - 2 758

View Full Product Details