Gene Transcripts

Gene Symbol
Rnf208
Entrez Gene ID
68846
Gene Name
ring finger protein 208
Gene Aliases
1110061N23Rik
Chromosome Location
Chr. 2: 25242929 - 25244261 on Build GRCm38
UniGene ID
Mm.24777
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176834.2 NP_789804.2 1 1006
GenBank mRNA AK004344.1 - 1 230
BC030073.1 - 2 808 5254639

View Full Product Details