Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008231.4 | NP_032257.3 | 3, 4 | 607 | |
GenBank mRNA | AF251787.1 | - | 3, 4 | 520 | |
AK005359.1 | - | 3, 4 | 386 | ||
AK017863.1 | - | 3, 4 | 561 | ||
AK029475.1 | - | 3, 4 | 351 | ||
AK185514.1 | - | 2, 3 | 219 | ||
BC005713.1 | - | 3, 4 | 517 | 2609261 | |
BC021654.1 | - | 3, 4 | 535 | 4163897 | |
D63707.1 | - | 3, 4 | 361 |