Gene Transcripts

Gene Symbol
Olfr263
Entrez Gene ID
18341
Gene Name
olfactory receptor 263
Gene Aliases
GA_x5J8B7TTCWC-858-415, IA3, MOR256-31, MOR256-37P, MOR256-61, Olfr263-ps1, Olfr42
Chromosome Location
Chr. 13: 21132777 - 21133730 on Build GRCm38
UniGene ID
Mm.246536
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010984.1 NP_035114.1 1 935
GenBank mRNA BC116918.1 - 1 1014
BC120698.1 - 1 1014 40129621

View Full Product Details