Gene Transcripts

Gene Symbol
Olfr1309
Entrez Gene ID
258439
Gene Name
olfactory receptor 1309
Gene Aliases
MOR245-5
Chromosome Location
Chr. 2: 111983134 - 111984096 on Build GRCm38
UniGene ID
Mm.377502
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146447.1 NP_666658.1 1 828
GenBank mRNA BC128010.1 - 1 832

View Full Product Details