Gene Transcripts

Gene Symbol
Ccnc
Entrez Gene ID
114839
Gene Name
cyclin C
Gene Aliases
Chromosome Location
Chr. 5: 35865600 - 35883732 on Build Rnor_6.0
UniGene ID
Rn.106758
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100472.1 NP_001093942.1 2 214
XM_006237937.2 XP_006237999.1 2 298
GenBank mRNA BC161908.1 - 2 284
D14013.1 - 2 199

View Full Product Details