Gene Transcripts

Gene Symbol
Psmc1
Entrez Gene ID
117263
Gene Name
proteasome (prosome, macropain) 26S subunit, ATPase, 1
Gene Aliases
s4
Chromosome Location
Chr. 6: 124123283 - 124135644 on Build Rnor_6.0
UniGene ID
Rn.10526
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057123.1 NP_476464.1 6 634
GenBank mRNA BC063157.1 - 7 646
D50696.1 - 6 634
FQ209916.1 - 7 639

View Full Product Details