Gene Transcripts

Gene Symbol
Pex6
Entrez Gene ID
117265
Gene Name
peroxisomal biogenesis factor 6
Gene Aliases
Paf-2, Paf2
Chromosome Location
Chr. 9: 16568743 - 16580900 on Build Rnor_6.0
UniGene ID
Rn.10675
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057125.1 NP_476466.1 1 741
GenBank mRNA D63673.1 - 1 741

View Full Product Details