Gene Transcripts

Gene Symbol
Ccl11
Entrez Gene ID
29397
Gene Name
chemokine (C-C motif) ligand 11
Gene Aliases
Scya11
Chromosome Location
Chr. 10: 69434965 - 69439566 on Build Rnor_6.0
UniGene ID
Rn.10632
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019205.1 NP_062078.1 2 170
GenBank mRNA U96637.1 - 2 136
Y08358.1 - 2 170

View Full Product Details